Skip to Content
Merck
All Photos(2)

Key Documents

C5438

Sigma-Aldrich

8-(4-Chlorophenylthio)-guanosine 3′,5′-cyclic monophosphate sodium salt

≥95% (HPLC), powder

Synonym(s):

8-pCPT-cGMP, pCPT-cGMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14ClN5NaO7PS
CAS Number:
Molecular Weight:
509.79
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white

solubility

H2O: 25 mg/mL

storage temp.

−20°C

SMILES string

[Na+].NC1=Nc2c(nc(Sc3ccc(Cl)cc3)n2[C@@H]4O[C@@H]5COP([O-])(=O)O[C@H]5[C@H]4O)C(=O)N1

InChI

1S/C16H15ClN5O7PS.Na/c17-6-1-3-7(4-2-6)31-16-19-9-12(20-15(18)21-13(9)24)22(16)14-10(23)11-8(28-14)5-27-30(25,26)29-11;/h1-4,8,10-11,14,23H,5H2,(H,25,26)(H3,18,20,21,24);/q;+1/p-1/t8-,10-,11-,14-;/m1./s1

InChI key

REEQGIQRCDWDRA-ZBMQJGODSA-M

Related Categories

Application

8-(4-Chlorophenylthio)-guanosine 3′,5′-cyclic monophosphate sodium salt has been used to activate GMP-dependent protein kinase (PKG) in vascular smooth muscle cells (VSMCs).

Biochem/physiol Actions

Membrane-permeable analog of cGMP that does not affect cGMP-regulated phosphodiesterase. A more potent cGMP analog than 8-Br-cGMP due to greater membrane permeability and a higher resistance to hydrolysis by phosphodiesterase. Used as a selective activator of cGMP dependent protein kinase (PKG). Found to be a very potent cyclic nucleotide-gated channel agonist.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the PKA & PKG page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stainless steel ions stimulate increased thrombospondin-1-dependent TGF-beta activation by vascular smooth muscle cells: implications for in-stent restenosis
Pallero MA, et al.
Journal of Vascular Research, 47(4), 309-322 (2010)
J Y Wei et al.
Journal of molecular neuroscience : MN, 10(1), 53-64 (1998-05-20)
Cyclic nucleotide-gated (CNG) channels are expressed in many cell types in both the nervous system and nonexcitable tissues. In order to understand the roles of cGMP-gated channels, and to distinguish actions of cGMP mediated through CNG channels from those through
Richard B Thorpe et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 304(9), R734-R743 (2013-03-01)
Chronic hypoxia attenuates soluble guanylate cyclase-induced vasorelaxation in serotonin (5-HT)-contracted ovine carotid arteries. Because protein kinase G (PKG) mediates many effects of soluble guanylate cyclase activation through phosphorylation of multiple kinase targets in vascular smooth muscle, we tested the hypothesis
M Suhasini et al.
Molecular and cellular biology, 18(12), 6983-6994 (1998-11-20)
Agents which increase the intracellular cyclic GMP (cGMP) concentration and cGMP analogs inhibit cell growth in several different cell types, but it is not known which of the intracellular target proteins of cGMP is (are) responsible for the growth-suppressive effects
P He et al.
The American journal of physiology, 274(6 Pt 2), H1865-H1874 (1998-06-25)
To investigate the mechanisms whereby guanosine 3',5'-cyclic monophosphate (cGMP) modulates microvessel permeability in vivo, we measured changes in microvessel hydraulic conductivity (Lp) and endothelial cytoplasmic Ca2+ concentration ([Ca2+]i) in response to the cGMP analogs 8-bromo-cGMP (8-BrcGMP) and 8-(p-chlorophenylthio)cGMP (8-pCPT-cGMP) in

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service