Skip to Content
Merck
All Photos(2)

Key Documents

C5105

Sigma-Aldrich

Anti-Calcium Channel (α2/δ-1 Subunit) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-CACNA2, Anti-CACNL2A, Anti-CCHL2A, Anti-LINC01112, Anti-lncRNA-N3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse, rat, pig, human

technique(s)

western blot (chemiluminescent): 1:200 using rat brain membranes

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Related Categories

General description

CACNA2D1 (calcium channel, voltage-dependent, α2/δ subunit 1) is a member of a family of GPI-anchored voltage-gated calcium channels (VGCCs)-associated subunits called α2δ. VGCC are composed of three types of subunits- pore-forming α1 and auxiliary β and α2δ subunits.

Immunogen

synthetic peptide corresponding to amino acids 1-15 of α2 subunit (amino acids 27-41 of rabbit α2/δ−1 precursor).

Application

Anti-Calcium Channel (α2/δ-1 Subunit) antibody produced in rabbit has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

CACNA2D1 (calcium channel, voltage-dependent, α2/δsubunit 1) is a type of α2δs, which are involved in the synaptic neurotransmitter release as they are concentrated at the synapses. They have a chaperone-like activity in non-neuronal cells. This protein acts as a target for gabapentin, which is a neuropathic analgesic. Mutations in this gene, in humans result in short QT syndrome (SQTS).

Target description

L-type of Voltage-gated Ca2+ Channel.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, and 0.05% sodium azide.

Preparation Note

Reconstitute the 0.05 mL vial with 50 μL deionized water. Reconstitute the 0.2 mL vial with 0.2 mL deionized water. After reconstitution, the antibody concentration is ~0.8 mg/mL.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kjara S Pilch et al.
The Journal of physiology, 600(24), 5333-5351 (2022-11-16)
In the mammalian brain, presynaptic CaV 2 channels play a pivotal role in synaptic transmission by mediating fast neurotransmitter exocytosis via influx of Ca2+ into the active zone of presynaptic terminals. However, the distribution and modulation of CaV 2.2 channels
Yuki Domon et al.
The Journal of pharmacology and experimental therapeutics, 365(3), 573-582 (2018-03-23)
Mirogabalin ([(1R,5S,6S)-6-(aminomethyl)-3-ethylbicyclo[3.2.0]hept-3-en-6-yl]acetic acid), a novel ligand for the α2δ subunit of voltage-gated calcium channels, is being developed to treat pain associated with diabetic peripheral neuropathy and postherpetic neuralgia. In the present study, we investigated the in vitro binding characteristics and
Kosuke Matsuda et al.
Frontiers in pharmacology, 15, 1382281-1382281 (2024-07-11)
Introduction: Atopic dermatitis (AD) is one of the most prevalent intractable chronic itch diseases worldwide. In recent years, new molecular-targeted drugs have emerged, but side effects and economic challenges remain. Therefore, since it is important for AD patients to have
Macroglia-derived thrombospondin 2 regulates alterations of presynaptic proteins of retinal neurons following elevated hydrostatic pressure
Wang S, et al.
Testing, 12(9), e0185388-e0185388 (2017)
Michael B Hoppa et al.
Nature, 486(7401), 122-125 (2012-06-09)
Synaptic neurotransmitter release is driven by Ca(2+) influx through active zone voltage-gated calcium channels (VGCCs). Control of active zone VGCC abundance and function remains poorly understood. Here we show that a trafficking step probably sets synaptic VGCC levels in rats

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service