Skip to Content
Merck
All Photos(1)

Key Documents

C3062

Sigma-Aldrich

Anti-Cholera Toxin antibody produced in rabbit

whole antiserum

Synonym(s):

Cholera Toxin Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

technique(s)

Ouchterlony double diffusion: 1:16
dot blot: 1:20,000
indirect ELISA: 1:8,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Cholera toxin, the pathogenic agent of cholera, is made of two subunits, A (27 kDa) and B (12 kDa) assembled with the stoichiometry AB5. The B-subunit binds to specific receptors, the monosialogangliosides GM1, located in the membrane of intestinal epithelial cells. The A1 fragment of the A-subunit is translocated through the membrane of the host cell, where it catalyses the ADP-ribosylation of the Gsa regulatory component of the adenylate cyclase complex. The resulting increased level of cyclic AMP promotes a wide variety of actions, including the secretion of chloride ions in the case of intestinal epithelial cells.
The antiserum reacts versus Cholera toxin, but shows no reaction versus Staphylococcal enterotoxin A, Staphylococcal enterotoxin B and Pseudomonas exotoxin A (protein concentration: 50-500 ng/dot). The product has not been tested for neutralization potency against active Cholera toxin.

Immunogen

toxin from Vibrio cholerae

Application

Anti-Cholera Toxin antibody produced in rabbit has been used in:
  • western blotting
  • quantitative ganglioside-dependent enzyme-linked immunoassay (ELISA)
  • double immunodiffusion

Biochem/physiol Actions

Cholera toxin (CT), a multifunctional protein plays a role in the immune system. It possesses immunomodulatory, adjuvant properties and also acts as an anti-inflammatory agent. Its immunomodulatory properties can be utilized to treat several autoimmune disorders. CT can serve as one of the best model of a multifunctional protein.

Preparation Note

delipidized

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purified cholera toxin B subunit from transgenic tobacco plants possesses authentic antigenicity
Wang XG, et al.
Biotechnology and Bioengineering, 72(4), 490-494 (2001)
Christine A Pellino et al.
Journal of bacteriology, 198(11), 1621-1630 (2016-03-24)
Shiga toxin (Stx)-producing Escherichia coli (STEC) is a major cause of foodborne illness, including the life-threatening complication hemolytic-uremic syndrome. The German outbreak in 2011 resulted in nearly 4,000 cases of infection, with 54 deaths. Two forms of Stx, Stx1 and
Generation and characterization of a live attenuated enterotoxigenic Escherichia coli combination vaccine expressing six colonization factors and heat-labile toxin subunit B
Turner AK, et al.
Clinical and Vaccine Immunology : CVI, 18(12), 2128-2135 (2011)
Qiyao Wang et al.
Infection and immunity, 83(9), 3381-3395 (2015-06-10)
Diverse environmental stimuli and a complex network of regulatory factors are known to modulate expression of Vibrio cholerae's principal virulence factors. However, there is relatively little known about how metabolic factors impinge upon the pathogen's well-characterized cascade of transcription factors
Tatiana El Hage et al.
The FEBS journal, 274(10), 2614-2629 (2007-04-25)
Using the in situ liver model system, we have recently shown that, after cholera toxin binding to hepatic cells, cholera toxin accumulates in a low-density endosomal compartment, and then undergoes endosomal proteolysis by the aspartic acid protease cathepsin-D [Merlen C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service