Skip to Content
Merck
All Photos(1)

Key Documents

C2773

Sigma-Aldrich

Cholesterol 5α,6α-epoxide

Synonym(s):

5α,6α-Epoxycholestan-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
EC Number:
MDL number:
UNSPSC Code:
41141804
PubChem Substance ID:
NACRES:
NA.77

Assay

≥80%

Quality Level

form

powder

functional group

epoxy

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](O)C3)(C)[C@@]3(O4)[C@@H]4C2)([H])CC[C@@]5(C)[C@@]1([H])CC[C@]5([H])[C@]([H])(C)CCCC(C)C

InChI

1S/C27H46O2/c1-17(2)7-6-8-18(3)21-9-10-22-20-15-24-27(29-24)16-19(28)11-14-26(27,5)23(20)12-13-25(21,22)4/h17-24,28H,6-16H2,1-5H3/t18-,19+,20+,21-,22+,23+,24+,25-,26-,27+/m1/s1

InChI key

PRYIJAGAEJZDBO-ZEQHCUNVSA-N

Application

Cholesterol 5α, 6α-epoxide was incorporated in culture medium of human arterial endothelial cells to study oxysterol-induced toxicity.

Biochem/physiol Actions

Cholesterol 5α, 6α-epoxide is an oxysterol, a cholesterol derivative by auto-oxidation. Oxysterols are non-genomic regulators of cholesterol homeostasis. The biological effects include protein prenylation, apoptosis, modulation of sphingolipid metabolism and platelet aggregation. Oxysterols bind to liver X receptors, modulate cholesterol efflux and decrease the uptake of cholesterol by the cells.

Preparation Note

Cholesterol 5α, 6α-epoxide yields clear, colorless solution in chloroform at 50 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chisato Ishimaru et al.
Lipids, 43(4), 373-382 (2008-01-25)
This paper describes the inhibitory activities of cholesterol derivatives such as cholesterol, sodium cholesteryl sulfate, cholesteryl-5alpha, 6alpha-epoxide, cholesteryl chloride, cholesteryl bromide, and cholesteryl hemisuccinate (compounds 1-6, respectively) against DNA polymerase (pol), DNA topoisomerase (topo), and human cancer cell growth. Among
Anne Vejux et al.
Histochemistry and cell biology, 127(6), 609-624 (2007-01-18)
Oxysterols, mainly those oxidized at the C7 position, induce a complex mode of cell death exhibiting some characteristics of apoptosis associated with a rapid induction of lipid rich multilamellar cytoplasmic structures (myelin figures) observed in various pathologies including atherosclerosis. The
Sinéad Lordan et al.
Journal of biochemical and molecular toxicology, 21(6), 362-372 (2007-11-13)
Oxidized low-density lipoprotein contains many potentially proatherogenic molecules, including oxysterols, which have been shown to induce apoptosis in various cell lines. The aim of this study was to investigate the pathway of apoptosis induced by oxidized low-density lipoprotein and the
May-Thu Ma et al.
Neuroscience letters, 476(1), 36-41 (2010-04-13)
Increase in levels of oxysterols or cholesterol oxidation products have been detected in brain areas undergoing neuroinflammation after excitotoxic injury, and the present study was carried out to elucidate possible effects of these products on exocytosis in rat pheochromocytoma-12 (PC12)
Y C O'Callaghan et al.
Cell biology and toxicology, 17(2), 127-137 (2001-08-14)
The cytotoxicity of the oxysterols 25-hydroxycholesterol, 7beta-hydroxycholesterol, cholesterol-5alpha,6alphaepoxide, cholesterol-5beta,6beta-epoxide, 19-hydroxycholesterol and 7-ketocholesterol was examined in U937 cells, a human monocytic blood cell line. 7beta-Hydroxycholesterol, cholesterol-5beta,6beta-epoxide, and 7-ketocholesterol, at 30 micromol/L concentration, were found to be cytotoxic to this cell line

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service