Skip to Content
Merck
All Photos(1)

Key Documents

C2643

Sigma-Aldrich

Coenzyme A, oxidized lithium salt

≥85%

Synonym(s):

CoA-S-S-CoA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H70N14O32P6S2 · xLi+
CAS Number:
Molecular Weight:
1533.05 (free acid basis)
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Quality Level

Assay

≥85%

form

solid

storage temp.

−20°C

InChI

1S/C42H70N14O32P6S2/c1-41(2,15-81-93(75,76)87-91(71,72)79-13-21-29(85-89(65,66)67)27(59)39(83-21)55-19-53-25-33(43)49-17-51-35(25)55)31(61)37(63)47-7-5-23(57)45-9-11-95-96-12-10-46-24(58)6-8-48-38(64)32(62)42(3,4)16-82-94(77,78)88-92(73,74)80-14-22-30(86-90(68,69)70)28(60)40(84-22)56-20-54-26-34(44)50-18-52-36(26)56/h17-22,27-32,39-40,59-62H,5-16H2,1-4H3,(H,45,57)(H,46,58)(H,47,63)(H,48,64)(H,71,72)(H,73,74)(H,75,76)(H,77,78)(H2,43,49,51)(H2,44,50,52)(H2,65,66,67)(H2,68,69,70)/t21-,22-,27-,28-,29-,30-,31+,32+,39-,40-/m1/s1

InChI key

YAISMNQCMHVVLO-ODFVJXNFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Coenzyme A (CoA) is a ubiquitous cofactor present in all living organisms. It is synthesized by the enzymatic conjugation of cysteine, pantothenate (Vitamin B5), and adenosine triphosphate (ATP).

Application

Coenzyme A oxidized lithium salt has been used in the preparation of the coenzyme A persulfide (CoASSH) standard to quantify the CoASSH in Staphylococcus aureus strains by tandem mass spectrometry.

Biochem/physiol Actions

Coenzyme A (CoA) plays a role as an acyl-group carrier and carbonyl activator in several biochemical reactions like the tricarboxylic acid cycle and fatty acid metabolism. It acts as a co-factor for several enzymes and is involved in several reactions in intermediary metabolism. CoA contains a thiol group that enables it to take part in several biochemical reactions. Oxidized CoA may be used to study reduction systems (CoA disulfide reductase (CoADR) systems) that regenerate reduced CoA in vivo. Furthermore, oxidized CoA may be used to study the unique functions of this molecule in vivo.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jamie R Wallen et al.
Biochemistry, 47(18), 5182-5193 (2008-04-11)
We have recently reported that CoASH is the major low-molecular weight thiol in Bacillus anthracis [Nicely, N. I. , Parsonage, D., Paige, C., Newton, G. L., Fahey, R. C., Leonardi, R., Jackowski, S., Mallett, T. C., and Claiborne, A. (2007)
Farhad Forouhar et al.
The Journal of biological chemistry, 280(48), 40328-40336 (2005-10-08)
Bacillus subtilis PaiA has been implicated in the negative control of sporulation as well as production of degradative enzymes. PaiA shares recognizable sequence homology with N-acetyltransferases, including those that can acetylate spermidine/spermine substrates. We have determined the crystal structure of
Ivan Gout
Biochemical Society transactions, 47(1), 469-476 (2019-02-21)
Coenzyme A (CoA) is an indispensable cofactor in all living organisms. It is synthesized in an evolutionarily conserved pathway by enzymatic conjugation of cysteine, pantothenate (Vitamin B5), and ATP. This unique chemical structure allows CoA to employ its highly reactive
Dennis R Harris et al.
The FEBS journal, 272(5), 1189-1200 (2005-02-22)
We have cloned NADH oxidase homologues from Pyrococcus horikoshii and P. furiosus, and purified the recombinant form of the P. horikoshii enzyme to homogeneity from Escherichia coli. Both enzymes (previously referred to as NOX2) have been shown to act as
J Luba et al.
Biochemistry, 38(9), 2725-2737 (1999-03-03)
An unusual flavoprotein disulfide reductase, which catalyzes the NADPH-dependent reduction of CoASSCoA, has recently been purified from the human pathogen Staphylococcus aureus [delCardayré, S. B., Stock, K. P., Newton, G. L., Fahey, R. C., and Davies, J. E. (1998) J.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service