Skip to Content
Merck
All Photos(1)

Key Documents

C2276

Sigma-Aldrich

Z-Gly-Pro-Arg p-nitroanilide acetate salt

protease substrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34N8O7 · xC2H4O2
CAS Number:
Molecular Weight:
582.61 (free base basis)
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

methanol: 20 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

CC(O)=O.NC(=N)NCCC[C@H](NC(=O)[C@@H]1CCCN1C(=O)CNC(=O)OCc2ccccc2)C(=O)Nc3ccc(cc3)[N+]([O-])=O

InChI

1S/C27H34N8O7.C2H4O2/c28-26(29)30-14-4-8-21(24(37)32-19-10-12-20(13-11-19)35(40)41)33-25(38)22-9-5-15-34(22)23(36)16-31-27(39)42-17-18-6-2-1-3-7-18;1-2(3)4/h1-3,6-7,10-13,21-22H,4-5,8-9,14-17H2,(H,31,39)(H,32,37)(H,33,38)(H4,28,29,30);1H3,(H,3,4)/t21-,22-;/m0./s1

InChI key

FPMAXHHYPVIJMC-VROPFNGYSA-N

Application

Z-Gly-Pro-Arg p-nitroanilide acetate salt has been used as a substrate in prostate-specific antigen (PSA) activation protease assay for determining enterokinase-PSA-trypsin (EK-PSA-TRP) activity and to perform kinetic experiments. It has also been used as a substrate to measure the pancreatic trypsin activity chromogenically.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver C Rogers et al.
Oncotarget, 9(32), 22436-22450 (2018-06-02)
Prostate cancer is the most diagnosed malignancy and the second leading cause of cancer-related death in American men. While localized therapy is highly curative, treatments for metastatic prostate cancer are largely palliative. Thus, new innovative therapies are needed to target
Constanze H Kubisch et al.
American journal of physiology. Gastrointestinal and liver physiology, 291(2), G238-G245 (2006-04-01)
Endoplasmic reticulum (ER) stress mechanisms have been found to play critical roles in a number of diseases states, such as diabetes mellitus and Alzheimer disease, but whether they are involved in acute pancreatitis is unknown. Here we show for the
Constanze H Kubisch et al.
American journal of physiology. Gastrointestinal and liver physiology, 291(2), G238-G245 (2006-04-01)
Endoplasmic reticulum (ER) stress mechanisms have been found to play critical roles in a number of diseases states, such as diabetes mellitus and Alzheimer disease, but whether they are involved in acute pancreatitis is unknown. Here we show for the
Jonathan M Bisaillon et al.
American journal of physiology. Cell physiology, 298(5), C993-1005 (2010-01-29)
We recently demonstrated that thapsigargin-induced passive store depletion activates Ca(2+) entry in vascular smooth muscle cells (VSMC) through stromal interaction molecule 1 (STIM1)/Orai1, independently of transient receptor potential canonical (TRPC) channels. However, under physiological stimulations, despite the ubiquitous depletion of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service