Skip to Content
Merck
All Photos(1)

Key Documents

C1235

Sigma-Aldrich

Cholesterol Oxidase microbial

recombinant, lyophilized powder, ≥10 units/mg protein

Synonym(s):

Cholesterol: oxygen oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

Quality Level

form

lyophilized powder

specific activity

≥10 units/mg protein

mol wt

55 kDa

solubility

50 mM potassium phosphate buffer, pH 7.0: soluble

storage temp.

−20°C

Application

Cholesterol oxidase is used to determine serum cholesterol. The enzyme also finds application in the microanalysis of steroids in food samples and in distinguishing 3-ketosteroids from 3b-hydroxysteroids. Transgenic plants expressing cholesterol oxidase are being investigated in the fight against the cotton boll weevil. CHOD has also been used as a molecular probe to elucidate cellular membrane structures.

Biochem/physiol Actions

Cholesterol oxidase (CHOD) is a monomeric flavoprotein containing FAD that catalyzes the first step in cholesterol catabolism. This bifunctional enzyme oxidizes cholesterol to cholest-5-en-3-one in an FAD-requiring step. This is subsequently isomerized to cholest-4-en-3-one with the release of H2O2. Optimum pH of the enzyme is 7.0. Hg2+, Ag+, ionic detergents inhibit the enzyme activity.

Unit Definition

One unit will convert 1.0 μmol of cholesterol to 4-cholesten-3-one per minute at 37 °C and pH 7.0 in a peroxidase linked system.

Preparation Note

Dissolves in cold 50 mM potassium phosphate buffer, pH 7.0. Solution is to be prepared just before use.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruno M Castro et al.
The Journal of biological chemistry, 284(34), 22978-22987 (2009-06-13)
A uniquely sensitive method for ceramide domain detection allowed us to study in detail cholesterol-ceramide interactions in lipid bilayers with low (physiological) ceramide concentrations, ranging from low or no cholesterol (a situation similar to intracellular membranes, such as endoplasmic reticulum)
Christophe A Marquette et al.
Analytical and bioanalytical chemistry, 390(1), 155-168 (2007-10-03)
The present review draws a general picture of the bioanalytical applications of electro-chemiluminescent reactions (ECL). Only the two main ECL reactions-i.e. the luminol-based and Ru(bpy)(3)(2+)-based reactions-are considered for application in the fields of enzyme biosensors, immunochemical biosensors, DNA biosensors, and
Porntip H Lolekha et al.
Clinica chimica acta; international journal of clinical chemistry, 339(1-2), 135-145 (2003-12-23)
Cholesterol oxidase is used for the determination of serum cholesterol. It can be derived from Streptomyces, Pseudomonas fluorescens, Cellulomonas, and Brevibacterium. This study compared the performance characteristics of four enzymes in the endpoint cholesterol determination. Using the Mega analyzer, we
Kwang-wook Ahn et al.
Biochemistry, 43(3), 827-836 (2004-01-21)
We investigated the dependence of cholesterol oxidase catalytic activity and membrane affinity on lipid structure in model membrane bilayers. The binding affinities of cholesterol oxidase to 100-nm unilamellar vesicles composed of mixtures of DOPC or DPPC and cholesterol are not
Effect of cholesterol concentration on organization of viral and vesicle membranes. Probed by accessibility to cholesterol oxidase.
R Pal et al.
The Journal of biological chemistry, 255(12), 5802-5806 (1980-06-25)

Protocols

Enzymatic activity assay for cholesterol oxidase, excluding specific product numbers, providing guidelines for accurate cholesterol oxidase assays.

Enzymatic activity assay for cholesterol oxidase, excluding specific product numbers, providing guidelines for accurate cholesterol oxidase assays.

Enzymatic activity assay for cholesterol oxidase, excluding specific product numbers, providing guidelines for accurate cholesterol oxidase assays.

Enzymatic activity assay for cholesterol oxidase, excluding specific product numbers, providing guidelines for accurate cholesterol oxidase assays.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service