Skip to Content
Merck
All Photos(1)

Key Documents

C1036

Sigma-Aldrich

Anti-Chicken Serum antibody produced in rabbit

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

chicken

technique(s)

immunoelectrophoresis: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Specificity

Strong reactivity with normal chicken serum has been determined by immunoelectrophoresis (IEP). This antiserum has not been assayed for interspecies cross reactivity.

Application

Anti-Chicken Serum antibody produced in rabbit has been used in enzyme-linked immunosorbent assay (ELISA) and immunoelectrophoresis.

Biochem/physiol Actions

Chicken serum may be suitable as medium supplement like fetal calf serum. It favors growth of parasite cultures providing all nutritional requirements It is employed for in vitro cultivation of L. infantum promastigotes.

Physical form

Rabbit Anti-Chicken Serum is provided as a liquid containing 0.1% sodium azide as preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of chicken (Gallus gallus) serum as a costly replacement for the fetal calf serum in cultivation of promastigotes of Leishmania infantum
Nasiri V, et al.
Asian Pacific Journal of Allergy and Immunology / Launched by the Allergy and Immunology Society of Thailand, 3(3), 169-173 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service