BMSW110512500
Accuris™ M1 Grade Calibration Weight Set
pack of 12 pieces, size 1-500 g, stainless steel
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.71
Recommended Products
material
stainless steel
packaging
pack of 12 pieces
manufacturer/tradename
Benchmark Scientific W1105-12500
size
1-500 g
Related Categories
General description
Basic, compact balances with readability in the range of 0.1g to 0.01g can be calibrated with M1 Class weights.
Each individual weight and weight set from Accuris™ is supplied with an individual Certificate of Verification confirming that the weight(s) meet OIML regulations and tolerances for the specific weight class.
Each individual weight and weight set from Accuris™ is supplied with an individual Certificate of Verification confirming that the weight(s) meet OIML regulations and tolerances for the specific weight class.
The economy M1 Calibration Weight Set includes 12 pieces: 1 x 1g, 2x2g, 1x5g, 1x10g, 2x20g, 1x50g, 1x100g, 2x200g, and 1x500g. A protective carrying case is included.
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)