BMSW1005100
Accuris™ M1 Grade Calibration Weight
size 100 g, stainless steel
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.71
Recommended Products
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific W1005-100
size
100 g
Related Categories
General description
- F1 Grade tolerance per OIML (International Organization of Legal Metrology)
- Works with all Accuris Lab balances
- Non-Magnetic Stainless Steel
- Includes a protective plastic case
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)