BAF367401023
Bel-Art® Earth Friendly Spoon
Wide Handle, capacity 5.0 mL
Synonym(s):
sampling spoon
All Photos(1)
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
Recommended Products
material
(PLA (polylactic acid) resin )
natural
packaging
package of 10
manufacturer/tradename
Bel-Art F36740-1023
capacity
5.0 mL
General description
Lessen environmental impact with these PLA (polylactic acid) resin sampling spoons. The production of 100% plant-based PLA contributes less greenhouse gasses and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource. Disposal options include industrial composting for biodegradability, incineration, recycling and landfill. Use a separate spoon for each material sampled to avoid cross contamination.
Features and Benefits
Green alternative to petroleum-based sampling tools; helps meet sustainability goals
Comparable in strength and durability to polystyrene, polypropylene and other plastic resin sampling tools
Comparable in strength and durability to polystyrene, polypropylene and other plastic resin sampling tools
Other Notes
Wide handles let you grip firmly and provide just enough stiffness to easily sample packed powders and other firm materials
Unique flat bottom shape allows convenient duo use as weighing boats to save time and assure accuracy
Unique flat bottom shape allows convenient duo use as weighing boats to save time and assure accuracy
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP