B9907
Nalgene® bottles, style 2105
capacity 250 mL
Synonym(s):
nalgene bottle, nalgene bottles
About This Item
Recommended Products
material
polypropylene cap
polypropylene copolymer bottles (PPCO)
round translucent bottle
Joint
Joints threaded neck (43)
description
Autoclavable
feature
with cap
packaging
pack of 12 ea
manufacturer/tradename
Nalgene® 2105-0008
bottle diam. × H
61 mm × 131 mm
capacity
250 mL
Related Categories
General description
Features and Benefits
- Nalgene® bottles are ideal for storing chemicals, materials, and specimens.
- Laboratory-grade PPCO material for reliably low leachables and extractables.
- Translucent - for visualization of samples.
- A wide neck enables easy transferring of contents.
- Bottles offer excellent durability and are chemically resistant to most acids, bases, and alcohols.
- Linerless caps ensure a leak-proof seal for sample protection.
- Autoclavable - The bottles can withstand temperatures up to +121°C.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
Related Content
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.