Skip to Content
Merck
All Photos(2)

Key Documents

B9139

Sigma-Aldrich

Biotin-dextran

mol wt 10,000, Lysine-fixable

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
eCl@ss:
34058011
NACRES:
NA.25

conjugate

biotin conjugate

Quality Level

form

powder

mol wt

10,000

extent of labeling

~2 mol per mol

color

white

solubility

H2O: soluble 50 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Biotin-dextran is used in tissue staining to prepare neurons for detection by avidin-conjugated detection systems. It is taken up by neuronal processes and transported bi-directionally. It infiltrates axon boutons in the anterograde direction and dendritic processes in the retrograde direction. Staining can be observed in fixed and sectioned tissue from two days to two weeks after biotin-dextran is injected into the brain or spinal cord. It can also be applied to cut nerve tracts. Biotinylated neurons are detected by either light or electron microscopy following incubation with avidin-horseradish peroxidase conjugate and the electron-dense substrate 3,3′-diaminobenzidine (DAB).

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shih-Yen Tsai et al.
Stroke, 42(1), 186-190 (2010-11-23)
we have shown that anti-Nogo-A immunotherapy to neutralize the neurite growth inhibitory protein Nogo-A results in functional improvement and enhanced plasticity after ischemic stroke in the adult rat. The present study investigated whether functional improvement and neuronal plasticity can be
Victoria L Cressman et al.
The Journal of comparative neurology, 518(14), 2693-2709 (2010-05-28)
Transformations in affective and social behaviors, many of which involve amygdalar circuits, are hallmarks of adolescence in many mammalian species. In this study, using the rat as a model, we provide the first evidence that afferents of the basal amygdala
Jia Liu et al.
Journal of the neurological sciences, 325(1-2), 127-136 (2013-01-16)
The stem cell-based experimental therapies are partially successful for the recovery of spinal cord injury (SCI). Recently, acellular spinal cord (ASC) scaffolds which mimic native extracellular matrix (ECM) have been successfully prepared. This study aimed at investigating whether the spinal
Bao-Ling Du et al.
Brain research, 1400, 87-98 (2011-06-11)
Delivery of cellular and/or trophic factors to the site of injury may promote neural repair or axonal regeneration and return of function after spinal cord injury. Engineered scaffolds provide a platform to deliver therapeutic cells and neurotrophic molecules. To explore
Yang-Can Duan et al.
Pflugers Archiv : European journal of physiology, 459(1), 1-9 (2009-07-25)
This study is to determine whether the nucleus of solitary tract (NTS) is an important component of the central neurocircuitry of the cardiac sympathetic afferent reflex (CSAR) and whether the gamma-aminobutyric acid (GABA) in the NTS modulates the CSAR. Renal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service