Skip to Content
Merck
All Photos(1)

Key Documents

B8291

Sigma-Aldrich

BSK-H Medium, Complete

With 6% rabbit serum, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Barbour-Stoenner-Kelly medium, Borrelia burgdorferi medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

product name

BSK-H Medium, Complete, sterile-filtered, with 6% rabbit serum, suitable for Borrelia burgdorferi

Quality Level

sterility

sterile-filtered

form

liquid

suitability

suitable for Borrelia burgdorferi

application(s)

food and beverages

components

NaHCO3: 2.068 g/L
phenol red: 0.01997 g/L
glucose: 5.64 g/L (Dextro)
HEPES: 5.64 g/L

shipped in

dry ice

storage temp.

−20°C

Application

Complete BSK-H Medium was used to culture B. burgdorferi to study the pathology of lyme borreliosis. It may be used in serum supplemented formulation for the growth and expansion of the Lyme desease spirochete, Borrelia burgdorferi.

Biochem/physiol Actions

BSK-H medium is a standardized complex medium designed to support the growth of the Lyme disease spirochete, Borrelia burgdorferi. To standardize the procedure for isolating and culturing Lyme disease spirochetes, Pollack, RJ et. al. modified the composition of the medium generally used for this purpose (BSK-II) and developed a system for its distribution. This medium contains no gelatin or agarose, and various components are used in proportions that differ from those in BSK-II. Each of the major proteinacious components was screened by substitution in samples of the complete product. The final medium was evaluated for the capacity to grow related spirochetes including Borrelia burgdorferi N40, Guilford, and JD-1 as well as strains of Borrelia hermsii (HS-1) and of Borrelia coriaceae (CO53). Standardized medium, supplemented with prescreened rabbit serum, facilitates comparison of research results between laboratories and may eventually permit definitive clinical diagnosis of Lyme disease based on demonstration of the pathogen. The standardized medium is designated BSK-H.
Barbour-Stonner-Kelly (BSK) medium is a modified medium for the culture of Borrelia strains that cause lyme borreliosis. BSK medium is a cheaper alternative for the selective culture of B. burgdorferi s.l. strains but not B. afzelii strain.

Preparation Note

BSK-H Medium is supplied as a sterile-filtered liquid. After thawing, mix well by inverting the bottle prior to use. If the thawed medium will not be used within a few days it is recommended that the medium be refrozen in working aliquots to avoid repeated free-thaw cycles. Other supplements may be added aseptically as desired. The nature of the supplement may affect storage conditions and shelf-life of the medium.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deborah A Grosenbaugh et al.
BMC veterinary research, 14(1), 312-312 (2018-10-18)
Prevention of Lyme disease in dogs in North America depends on effective vaccination against infection by the tick vector-born spirochete Borrelia burgdorferi. Most vaccines effectively prevent spirochete transmission to dogs during tick feeding based on immunization with the outer-surface lipoprotein
Ghiabe H Guibinga et al.
Human vaccines & immunotherapeutics, 16(9), 2114-2122 (2020-08-14)
Lyme disease is the most common vector-borne disease in North America. The etiological agent is the spirochete Borreliella burgdorferi, transmitted to mammalian hosts by the Ixodes tick. In recent years there has been an increase in the number of cases
Sin Hang Lee et al.
American journal of clinical pathology, 133(4), 569-576 (2010-03-17)
The DNA of Borrelia burgdorferi spirochetes extracted by ammonium hydroxide was used as the template for nested polymerase chain reaction (PCR) amplification of the species-specific 16S ribosomal DNA (rDNA). The primers were those well known to be specific for signature
Nataliia Rudenko et al.
Journal of clinical microbiology, 46(10), 3540-3543 (2008-07-25)
Molecular analysis of a clinical sample confirmed the presence of Borrelia bissettii DNA in cardiac valve tissue from a patient with endocarditis and aortic valve stenosis. This evidence strongly supports the involvement of B. bissettii in Lyme disease in Europe.
Jana Tuhácková et al.
Biomedical papers of the Medical Faculty of the University Palacky, Olomouc, Czechoslovakia, 149(2), 297-302 (2006-04-08)
Verification of the efficacy of Biocan B inj. ad us. vet. (Bioveta, a.s.) was done by challenge testing. Ticks collected in the nature were used as natural vectors of the infection. Six beagles and two control ones were used in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service