Skip to Content
Merck
All Photos(1)

Key Documents

B7786

Sigma-Aldrich

Anti-fd Bacteriophage antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-fd Bacteriophage

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

packaging

antibody small pack of 25 μL

technique(s)

indirect ELISA: 1:1,000 using 5 × 1010 phage/ml

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Specificity

The antibody binds specifically to phage coat proteins of fd phage or M13 phage and thus may act as a capture antibody when coated directly on multiwell plates or as a primary detection antibody for specifically captured fd or M13 phage. The antibody may be useful in rapidly sorting large phage display libraries (antibody, peptide, etc.) with the expressed proteins fused to either the gene III or to the gene VIII protein of the filamentous phage. The antibody may be used as a reagent in "phage ELISA" offering sensitive and specific activity for detection of recombinant phages.

Immunogen

fd bacteriophage

Application

Anti-fd Bacteriophage antibody produced in rabbit may be used in ELISA at a dilution of 1:1000. It was used to in antiphage immunohistochemistry.
B7786 Rabbit Anti-fd Bactieriophage may be used to sort phage display libraries (antibody, peptide, etc.) with expressed proteins fused to either the gene III or gene VIII protein of filamentous bacteriophage. The product may also be used in "phage ELISA" offering specific activity for detection of recombinant bacteriophage. It will also function as a capture antibody when coated directly on microwell plates or as primary detection antibody for specifically captured fd or M13 bacteriophage.

Physical form

Solution supplied in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Preparation Note

Anti-fd Bacteriophage is developed in rabbit using repeated injections of fd bacteriophage as the immunogen. Whole antiserum is fractionated and then further purified by ion-exchange chromatography to provide the IgG fraction of antiserum which is essentially free of other rabbit serum proteins.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuto Akiyama et al.
Biomedical research (Tokyo, Japan), 35(2), 105-116 (2014-04-25)
Of all potential biological therapeutics, monoclonal antibody (mAb)-based therapies are becoming the dominant focus of clinical research. In particular, smaller recombinant antibody fragments such as single-chain variable fragments (scFv) have become the subject of intense focus. However, an efficient affinity
Mikhail G Kolonin et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 20(7), 979-981 (2006-04-04)
In vivo phage display is a technology used to reveal organ-specific vascular ligand-receptor systems in animal models and, recently, in patients, and to validate them as potential therapy targets. Here, we devised an efficient approach to simultaneously screen phage display
Paladd Asavarut et al.
EMBO molecular medicine, 14(8), e15418-e15418 (2022-06-28)
Immunotherapy is a powerful tool for cancer treatment, but the pleiotropic nature of cytokines and immunological agents strongly limits clinical translation and safety. To address this unmet need, we designed and characterised a systemically targeted cytokine gene delivery system through
Mikhail G Kolonin et al.
Nature medicine, 10(6), 625-632 (2004-05-11)
Obesity is an increasingly prevalent human condition in developed societies. Despite major progress in the understanding of the molecular mechanisms leading to obesity, no safe and effective treatment has yet been found. Here, we report an antiobesity therapy based on
Paola A Bignone et al.
Methods in molecular biology (Clifton, N.J.), 1357, 269-283 (2014-11-21)
The ability of human pluripotent stem cells (hPS) to both self-renew and differentiate into virtually any cell type makes them a promising source of cells for cell-based regenerative therapies. However, stem cell identity, purity, and scalability remain formidable challenges that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service