B7410
Nalgene® bottles, style 2120
capacity 4,000 mL
Synonym(s):
HDPE bottles, nalgene bottle, nalgene bottles
About This Item
Recommended Products
material
high-density polyethylene bottle
polypropylene cap
polypropylene screw closure
round natural bottle
Joint
Joints threaded neck (100)
description
Not autoclavable
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
Nalgene® 2120-0010
bottle diam. × H
153 mm × 288 mm
capacity
4,000 mL
General description
Features and Benefits
- Versatile as it is chemical-resistant to acids, bases, and alcohols
- Excellent for freezer storage up to −100°C (−148°F) for long-term purposes
- Non-leakproof for liquids
- Made from non-toxic materials
- Linerless closures are recommended to prevent leaks, wrinkles, and contamination of reagents
Legal Information
Certificates of Analysis (COA)
Need A Sample COA?
This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.