Skip to Content
Merck
All Photos(1)

Key Documents

B7022

Sigma-Aldrich

Anti-Mouse IgG (γ-chain specific)−Biotin antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

biotin conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:20,000

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases . Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice . Goat Anti-Mouse IgG (γ-chain specific)-Biotin antibody specific for mouse IgG when tested against mouse IgA, IgG (all subclasses), and IgM.

Immunogen

Mouse IgG

Application

Anti-Mouse IgG (γ-chain specific)-Biotin antibody ELISA is suitable for use in ELISA (1:20,000) and direct immunomagnetic sorting (1:150 ).

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valeria Carolina Colombo et al.
Transboundary and emerging diseases (2021-07-06)
SARS-CoV-2 human-to-animal transmission can lead to the establishment of novel reservoirs and the evolution of new variants with the potential to start new outbreaks in humans. We tested Norway rats inhabiting the sewer system of Antwerp, Belgium, for the presence
Immunomagnetic separation of cells of the toxic dinoflagellate Alexandrium fundyense from natural plankton samples
Aguilera, A., et al.
Marine Ecology Progress Series, 143, 255-269 (1996)
Maria Victoria Sanchez et al.
PloS one, 9(8), e104824-e104824 (2014-08-21)
There is a critical need for new influenza vaccines able to protect against constantly emerging divergent virus strains. This will be sustained by the induction of vigorous cellular responses and humoral immunity capable of acting at the portal of entry
K M Lima et al.
Gene therapy, 10(8), 678-685 (2003-04-15)
The high incidence of tuberculosis around the world and the inability of BCG to protect certain populations clearly indicate that an improved vaccine against tuberculosis is needed. A single antigen, the mycobacterial heat shock protein hsp65, is sufficient to protect
Jun P Hong et al.
Cell reports. Medicine, 1(3) (2020-07-24)
CD137 is a costimulatory receptor expressed on natural killer cells, T cells, and subsets of dendritic cells. An agonistic monoclonal antibody (mAb) against CD137 has been used to reduce tumor burden or reverse autoimmunity in animal models and clinical trials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service