Skip to Content
Merck
All Photos(1)

Key Documents

B4563

Sigma-Aldrich

Bisantrene dihydrochloride

≥98% (HPLC)

Synonym(s):

Bis((4,5-dihydro-1H-imidazol-2-yl)hydrazone)-9,10-anthracenedicarboxaldehyde dihydrochloride, NSC 337766

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22N8 ·2HCl
CAS Number:
Molecular Weight:
471.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

solubility

deionized water: 8 mg/mL

storage temp.

room temp

SMILES string

Cl.Cl.C1CN=C(N1)N\N=C\c2c3ccccc3c(\C=N\NC4=NCCN4)c5ccccc25

InChI

1S/C22H22N8.2ClH/c1-2-6-16-15(5-1)19(13-27-29-21-23-9-10-24-21)17-7-3-4-8-18(17)20(16)14-28-30-22-25-11-12-26-22;;/h1-8,13-14H,9-12H2,(H2,23,24,29)(H2,25,26,30);2*1H/b27-13+,28-14+;;

InChI key

KINULKKPVJYRON-PVNXHVEDSA-N

Application

Bisantrene (NSC 337766), and its derivatives are topoisomerase II poisons and DNA intercalators. It may be used as model compounds to study P-glycoprotein-mediated multiple drug resistance (MDR1). Bisantrene may be used as a Rac1 inhibitor.

Biochem/physiol Actions

Bisantrene dihydrochloride is an antineoplastic agent, MDR1 substrate, DNA intercalator, and topoisomerase II poison. Cancer cells that develop resistance to bisantrene tend to overexpress P-glycoprotein. Bisantrene can be used to select for P-glycoprotein-mediated multiple drug resistance. The data suggest that bisantrene is an excellent substrate for P-glycoprotein.
Bisantrene is an anthracene bishydrazone derivative, which has antitumor activity.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Yao et al.
Science in China. Series B, Chemistry, life sciences & earth sciences, 38(12), 1462-1472 (1995-12-01)
The binding of bisantrene to four DNA tetramers, d(CGCG)2, d(GCGC)2, d(CATG)2, and d(GTAC)2, was investigated by 1D and 2D NMR spectroscopy. Bisantrene is a well known anticancer drug and has been used clinically for years. DNA is believed to be
G Visani et al.
Haematologica, 75(6), 527-531 (1990-11-01)
In vitro clonogenic assays may be useful for determining the sensitivity of leukemic cells to chemotherapeutic agents. We evaluated the antileukemic effect of Bisantrene (an anthracene derivative now undergoing phase II clinical trials in relapsed/resistant acute non lymphoid leukemias-ANLL) using
Disposition of bisantrene in humans and rabbits: evidence for intravascular deposition of drug as a cause of phlebitis
Powis G and Kovach JS
Cancer Research, 43(2), 925-929 (1983)
A Spadea et al.
Leukemia & lymphoma, 9(3), 217-220 (1993-02-01)
Because of the lack of standard treatment in refractory and relapsed acute myelogenous leukemia (AML) several new drugs have been employed alone to evaluate their efficacy in this peculiar category of patient. Bisantrene, a new anthracene bishydrazone derivative, has shown
K C Murdock et al.
Journal of medicinal chemistry, 36(15), 2098-2101 (1993-07-23)
The selective phosphorylation of bisantrene (1) affords bis(phosphonoguanidinic acid) 6, a prodrug with enhanced aqueous solubility (as sodium salt 7) at physiological pH. Unlike 1, in a rat tail vein model, no precipitation was observed when bis(phosphonoguanidinic acid) 6 was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service