Skip to Content
Merck
All Photos(2)

Key Documents

B4559

Sigma-Aldrich

BRL 44408 maleate salt

≥98% (HPLC)

Synonym(s):

2-[2H-(1-Methyl-1,3-dihydroisoindole)methyl]-4,5-dihydroimidazole maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N3O4
CAS Number:
Molecular Weight:
331.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: soluble 15 mg/mL, clear

originator

GlaxoSmithKline

SMILES string

OC(=O)\C=C/C(O)=O.CC1N(CC2=NCCN2)Cc3ccccc13

InChI

1S/C13H17N3.C4H4O4/c1-10-12-5-3-2-4-11(12)8-16(10)9-13-14-6-7-15-13;5-3(6)1-2-4(7)8/h2-5,10H,6-9H2,1H3,(H,14,15);1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

DDIQGSUEJOOQQQ-BTJKTKAUSA-N

Biochem/physiol Actions

BRL 44408 maleate is a selective α2A-adrenoceptor antagonist.

Features and Benefits

This compound is featured on the α2-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold under agreement from Glaxo­Smith­Kline

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baogang Liu et al.
Neuroreport, 17(12), 1313-1317 (2006-09-05)
Perineural injection of clonidine at the site of nerve injury reduces hypersensitivity while simultaneously reducing leukocyte number and cytokine expression and hyperexcitability in sensory neurons. The activation of p38 mitogen-activated protein kinase in sensory neurons contributes to the development and
Alfonso Romero-Sandoval et al.
Anesthesiology, 104(2), 351-355 (2006-01-27)
Partial sciatic nerve ligation (PSNL) produces axonal damage, a local inflammatory response, and wallerian degeneration. Cytokines secreted near the site of nerve injury are thought to play important roles in development and maintenance of central sensitization and neuropathic pain. Injection
Zoltán S Zádori et al.
Neurochemistry international, 51(5), 297-305 (2007-08-01)
The inhibitory effect of clonidine (non-selective alpha2-adrenoceptor agonist) and oxymetazoline (alpha2A-adrenoceptor selective agonist) was compared on basal and stimulated gastric motor activity (gastric tone and contractions) using the balloon method in the rat. It was shown that oxymetazoline (0.2-1.7 micromol/kg
Shawquia Young et al.
Basic & clinical pharmacology & toxicology, 107(2), 690-697 (2010-04-01)
Previously, we reported that antinociceptive synergism of a 5-HT(3)/alpha(2)-adrenoceptor ligand MD-354 (m-chlorophenylguanidine) and clonidine combination occurs, in part, through a 5-HT(3) receptor antagonist mechanism. In the present investigation, a possible role for alpha(2)-adrenoceptors was examined. Mechanistic studies using yohimbine (a
Roberto T Sudo et al.
The journal of pain : official journal of the American Pain Society, 11(1), 71-78 (2009-10-27)
Finding new chemicals or adjuvants with analgesic effects in the central nervous system is clinically relevant due to the limited number of drugs with these properties. Here, we present PT-31, which is chemically related to 3-benzyl-imidazolidine, with an analgesic profile

Articles

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service