Skip to Content
Merck
All Photos(1)

Key Documents

B4311

Sigma-Aldrich

BF-170 hydrochloride

≥98% (HPLC), solid

Synonym(s):

2-(4-aminophenyl)quinoline hydrochloride, NSC 168387 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2 · HCl
CAS Number:
Molecular Weight:
256.73
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated
protect from light
under inert gas

color

orange

solubility

H2O: 10 mg/mL

storage temp.

−20°C

SMILES string

Cl[H].Nc1ccc(cc1)-c2ccc3ccccc3n2

InChI

1S/C15H12N2.ClH/c16-13-8-5-12(6-9-13)15-10-7-11-3-1-2-4-14(11)17-15;/h1-10H,16H2;1H

InChI key

UYYPMQOFLJHJAW-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

BF-170 is a new probe for neurofibrillary tangles (tau fibrils) with greater binding affinity to tau than to Aβ fibrils.
BF-170 is a new probe for neurofibrillary tangles (tau fibrils). It exhibits greater binding affinity to tau than to Aβ fibrils, EC50 = 221 nM vs. EC50 = 786 nM (Ki for Aβ > 5,000 nM). BF-170 demonstrates fast clearance from the brain; clearly visualizes neurofibrillary tangles, neuropil threads, and paired helical filament-type neuritis. BF-70 may become one of the candidate compounds for in vivo imaging of tau pathology associated with Alzheimer′s disease and is a useful tool in distinguishing among tau and Aβ fibrils specifically in AD.

Caution

Product is air sensitive, store under inert gas.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service