Skip to Content
Merck
All Photos(1)

Key Documents

B154

Sigma-Aldrich

BU224 hydrochloride

solid

Synonym(s):

2-(4,5-Dihydroimidazol-2-yl)quinoline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N3 · HCl
Molecular Weight:
233.70
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

white

solubility

ethanol: 1.2 mg/mL
H2O: >16 mg/mL
0.1 M NaOH: 24 mg/mL
0.1 M HCl: >30 mg/mL

SMILES string

Cl[H].C1CN=C(N1)c2ccc3ccccc3n2

InChI

1S/C12H11N3.ClH/c1-2-4-10-9(3-1)5-6-11(15-10)12-13-7-8-14-12;/h1-6H,7-8H2,(H,13,14);1H

InChI key

DDFHQXAQWZWRSQ-UHFFFAOYSA-N

Biochem/physiol Actions

I2 Imidazoline receptor antagonist.

Features and Benefits

This compound is featured on the Imidazoline Binding Sites page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Hygroscopic; store desiccated

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Diaz et al.
European journal of pharmacology, 333(1), 9-15 (1997-08-20)
In this electrophysiological study, the effect of BU-224 (2-(4,5-dihydroimidazol-2yl)-quinoline hydrochloride)), a novel high affinity imidazoline I2 receptor ligand, was tested on the responses of nociceptive neurones in the spinal dorsal horn. When applied spinally, akin to an intrathecal application (i.t.)
A L Hudson et al.
Annals of the New York Academy of Sciences, 881, 81-91 (1999-07-23)
Over several years our group has sought to synthesize and identify selective ligands for imidazoline (I) receptors, in particular the I2 binding site. As a consequence, [3H]2-(2-benzofuranyl)-2-imidazoline (2BFI) has proved extremely useful for binding and autoradiographic studies. More recently we
Kazunori Kawamura et al.
Nuclear medicine and biology, 45, 1-7 (2016-11-12)
A positron emission tomography (PET) probe with ultra-high specific radioactivity (SA) enables measuring high receptor specific binding in brain regions by avoiding mass effect of the PET probe itself. It has been reported that PET probe with ultra-high SA can
Eloísa Rubio-Beltrán et al.
European journal of pharmacology, 812, 97-103 (2017-07-12)
Calcitonin gene-related peptide (CGRP) plays a role in several (patho)physiological functions, and modulation of its release is considered a therapeutic target. In this respect, electrical spinal (T
Laura Micheli et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 17(3), 1005-1015 (2020-06-24)
Anti-cancer therapy based on the repeated administration of oxaliplatin is limited by the development of a disabling neuropathic syndrome with detrimental effects on the patient's quality of life. The lack of effective pharmacological approaches calls for the identification of innovative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service