Skip to Content
Merck
All Photos(1)

Key Documents

B1439

Sigma-Aldrich

Brassinolide

≥80% (TLC)

Synonym(s):

(1R,3aS,3bS,6aS,8S,9R,10aR,10bS,12aS)-1-[(1S,2R,3R,4S)-2,3-Dihydroxy-1,4,5-trimethylhexyl]hexadecahydro-8,9-dihydroxy-10a,12a-dimethyl-6H-benz[c]indeno[5,4-e]oxepin-6-one, (2a,3a,5a,22R,23R,24S)-2,3,22,23-Tetrahydroxy-B-homo-7-oxaergostan-6-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H48O6
Molecular Weight:
480.68
UNSPSC Code:
10171502
NACRES:
NA.25

Quality Level

Assay

≥80% (TLC)

form

powder

color

white to off-white

storage temp.

room temp

General description

Brassinolide is a plant growth hormone and the most biologically active brassinosteroid (BR). It has been implicated in Auxin signaling. It has also been used for studying plant biosynthetic pathways. Brassinolide might also regulate water-transport properties of cell membranes in Arabidopsis thaliana.

Application

Brassinolide has been used as a plant growth hormone.

Biochem/physiol Actions

Brassinolide is a plant growth hormone that has been implicated in Auxin signaling. It has also been used for studying plant biosynthetic pathways.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biosynthetic pathways of brassinolide in Arabidopsis.
Noguchi T, et al.
Plant Physiology, 124(1), 201-210 (2000)
Brassinolide may control aquaporin activities in Arabidopsis thaliana.
Morillon R, et al.
Planta, 212(2), 199-204 (2001)
Zongqi Xu et al.
Plant physiology and biochemistry : PPB, 118, 460-470 (2017-07-26)
Poly-γ-glutamic acid (γ-PGA) is a microbe-secreted isopeptide shown to promote growth and enhance crop stress tolerance. However, its downstream signaling pathways are unknown. Here, we studied γ-PGA-induced tolerance to salt and cold stresses. Pretreatment with γ-PGA contributed to enhance stress
Optimization of Phenotyping Assays for the Model Monocot
Biswa R Acharya et al.
Frontiers in plant science, 8, 2172-2172 (2018-01-10)
Chang-Jie Jiang et al.
Frontiers in plant science, 8, 1558-1558 (2017-09-22)
In response to pathogen attack, plants prioritize defense reactions generally at the expense of plant growth. In this work, we report that changes in phytohormone signaling pathways are associated with the stunted plant growth caused by blast disease in rice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service