Skip to Content
Merck
All Photos(2)

Key Documents

B0681

Sigma-Aldrich

Anti-BACE 1, N-Terminus (46-62) antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-β-Site APP Cleaving Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 60-75 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

western blot: 1:1,000 using a whole cell extract from the human kidney HEK293 cell line stably transfected with human BACE-1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BACE1(23621)

General description

The membrane-associated aspartic protease BACE-1 (β-site APP cleaving enzyme, Asp2 or memapsin 2) has been identified as β-secretase. BACE-1 constitutes the predominant β-secretase activity in human brain tissue. It is highly expressed in neurons, the major site of Aβ generation. BACE-1 is localized within the Golgi and endosomal compartments, among the several intracellular sites where Aβ is thought to be produced. BACE-1 is initially an inactive proenzyme and localized in endoplasmic reticulum.

Immunogen

synthetic peptide corresponding to the N-terminus of human BACE-1 (amino acids 46-62).

Application

Anti-BACE 1, N-Terminus (46-62) antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Overexpression of β-site APP cleaving enzyme, Asp2 or memapsin 2 (BACE-1) leads to increased β-secretase activity while displaying appropriate cleavage site specificity for APP. The N-glycosylation and phosphorylation of BACE-1 within its C-terminal domain regulated its intracellular trafficking.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activity-induced convergence of APP and BACE-1 in acidic microdomains via an endocytosis-dependent pathway
Das U, et al.
Neuron, 79(3), 447-460 (2013)
BACE1 SUMOylation increases its stability and escalates the protease activity in Alzheimer?s disease
Bao J, et al.
Proceedings of the National Academy of Sciences of the USA, 115(15), 3954-3959 (2018)
Latha Devi et al.
Current Alzheimer research, 12(1), 13-21 (2014-12-20)
The β-secretase enzyme BACE1, which initiates the cleavage of amyloid precursor protein (APP) into the amyloid-β (Aβ) peptide, is a prime therapeutic target for Alzheimer's disease (AD). However, recent investigations using genetic animal models raise concern that therapeutic BACE1 inhibition
BACE1 protein endocytosis and trafficking are differentially regulated by ubiquitination at lysine 501 and the Di-leucine motif in the carboxyl terminus
Kang EL, et al.
The Journal of Biological Chemistry, 287(51), 42867-42880 (2012)
L Devi et al.
Neuroscience, 307, 128-137 (2015-09-01)
β-Site APP-cleaving enzyme 1 (BACE1) initiates the generation of amyloid-β (Aβ), thus representing a prime therapeutic target for Alzheimer's disease (AD). Previous work including ours has used BACE1 haploinsufficiency (BACE1(+/-); i.e., 50% reduction) as a therapeutic relevant model to evaluate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service