AXYTXL10LRS
MAXYMum Recovery® pipette tips, for Pipetman® P2/P10 and Eppendorf® Research/Reference pipettors, ultramicro, extra long
size 0.1-10 μL, pkg of 5x960tips/cs, racked, clear, sterile
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
size:
0.1-10 μL
sterility:
sterile
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc TXL-10-L-R-S
Corning
Corning
Recommended Products
sterility
sterile
manufacturer/tradename
Axygen Inc TXL-10-L-R-S
Corning
packaging
pkg of 5x960tips/cs
size
0.1-10 μL
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
Other Notes
Browse all pipette tips
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Maxymum Recovery is a registered trademark of Corning, Inc.
Pipetman is a registered trademark of Gilson S.A.S. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service