AXYTF400RS
Pipette filter tips
for Eppendorf® pipettors, ultramicro, racked, size 0.5-10 μL, pkg of 5x960tips/cs, sterile
Synonym(s):
Pipette tips, pipette tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121601
NACRES:
NB.02
size:
0.5-10 μL
sterility:
sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter
RNase free
barrier: no
endotoxin free
filter
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc TF-400-R-S
Corning
Corning
Recommended Products
material
clear
sterility
sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter
manufacturer/tradename
Axygen Inc TF-400-R-S
Corning
packaging
pkg of 5x960tips/cs
size
0.5-10 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Axygen® Filter Barrier tips can be used for DNA amplification and microbiological applications where slight contamination can alter the results.
Features and Benefits
Axygen® Filter Barrier tips have the following features and benefits:
- Tips create a physical barrier to prevent cross-contamination and block aerosols
- The DNA inhibitors free-polyethylene filters are fit into the tips robotically to avoid nuclease contamination
- Packed in sealed-bottom racks with lids
- Tips are RNAse and DNAse free
- Non-pyrogenic
Other Notes
Browse all pipette tips
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service