Skip to Content
Merck
All Photos(1)

Key Documents

AXYSCT200CS

Screw cap tube

size 2.0 mL, clear, pkg of 8x500caps/cs, sterile

Synonym(s):

centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

material

conical bottom tube
polypropylene
screw top cap

sterility

sterile

manufacturer/tradename

Axygen Inc SCT-200-C-S
Corning

packaging

pkg of 8x500caps/cs

parameter

20000 × g max. RCF

size

2.0 mL

color

clear
clear

fitting

cap (screw cap)

suitability

suitable for (centrifugation)

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service