Skip to Content
Merck
All Photos(1)

Key Documents

AXYMCT150B

Boil proof microtubes

size 1.5 mL, blue, pkg of 10x500tubes/cs

Synonym(s):

centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702
NACRES:
NB.22

material

cap (snap cap)
conical bottom tube
polypropylene
polypropylene cap

sterility

non-sterile

feature

RNase and DNase free
endotoxin free
graduations

manufacturer/tradename

Axygen Inc MCT-150-B
Corning

packaging

pkg of 10x500tubes/cs

size

1.5 mL

color

blue

fitting

cap (push fit)

suitability

suitable for (centrifugation)

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service