Skip to Content
Merck
All Photos(1)

Key Documents

AXYGD1000

Corning® Axygen® Gel Documentation System

Synonym(s):

gel system accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

manufacturer/tradename

Corning GD1000

Looking for similar products? Visit Product Comparison Guide

General description

Corning®Axygen® Gel Documentation System is easy to set up and has an easy interface for contrast adjusting, annotation, and image capture that allows for the simple saving and opening of images for more thorough analysis without manipulating the camera manually.

Application

Corning® Axygen® Gel Documentation System has been used in the semi-quantitative detection of enterocytozoon hepatopenaei (EHP) DNA.

Features and Benefits

  • Auto exposure tool determines the ideal exposure time with just one click and the slider tool can be used manually to view an updated live image and adjust the exposure time
  • Only the selected region is used to calculate the auto exposure which is beneficial for gels with strong or weak bands
  • UV 302, UV 365, epi white, or epi blue light sources, as well as an optional trans white light illumination tray, are all selectable through the darkroom tab
  • An area of interest can be chosen using the select ROI tool
  • To view the gel with the lights on, the band excision tool temporarily disables the system′s safety measures
  • Crop, rotate, resize, contrast, saturation, and zoom are all imaging tools and text and drawing tools are included in annotation tools
  • For further analysis, it is possible to save, print, email, or export finished images
  • UV light is automatically turned off by a door sensor to prevent accidental exposure
  • Band cutting on the UV transilluminator is possible with safety override, and it turns off automatically after five minutes
  • Intel®processor, 1.4 GHz processor speed, 2GB RAM, 16GB free hard disk space, and 2 USBs are minimum hardware requirements
  • Epi White light sources with dual-wavelength transillumination at 302 nm are used
  • Compatible with Mircrosoft®Windows® 7 or higher operating system
  • When working with UV light, always wear protective eyewear

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Microsoft is a registered trademark of Microsoft Corporation
Pentium is a registered trademark of Intel Corp.
Windows is a registered trademark of Microsoft Corporation

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple filter paper-based method for transporting and storing Enterocytozoon hepatopenaei DNA from infected Litopenaeus vannamei tissues
Karthikeyan, Saranya and K
Journal of Invertebrate Pathology, 169, 107305-107305 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service