Skip to Content
Merck
All Photos(1)

Key Documents

AXYEV50R

Pipette tips for Tecan® EVO/ Caliper Zephyr® Automated Liquid Handling System

racked, capacity 50 μL, pkg of 5x960tips/cs, clear

Synonym(s):

Pipette tips for Velocity

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
capacity:
50 μL
sterility:
non-sterile
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc EV-50-R
Corning

sterility

non-sterile

manufacturer/tradename

Axygen Inc EV-50-R
Corning

packaging

pkg of 5x960tips/cs

capacity

50 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Tecan is a registered trademark of Tecan Group AG
Zephyr is a registered trademark of Caliper Life Sciences, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service