Skip to Content
Merck
All Photos(3)

Key Documents

AV51158

Sigma-Aldrich

Anti-APOH antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Apolipoprotein H (β-2-glycoprotein I), Anti-B2G1, Anti-BG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

human, dog, pig, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APOH(350)

Immunogen

Synthetic peptide directed towards the N terminal region of human APOH

Application

Anti-APOH antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/ml.

Biochem/physiol Actions

Apolipoprotein H (APOH; beta-2-glycoprotein I) is a phospholipid implicated in lipid metabolism, coagulation and production of antiphospholipid autoantibodies. It functions as a cofactor required for the binding of antiphospholipid antibodies present in the sera of lupus and antiphospholipid syndrome patients.

Sequence

Synthetic peptide located within the following region: LWPINTLKCTPRVCPFAGILENGAVRYTTFEYPNTISFSCNTGFYLNGAD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service