Skip to Content
Merck
All Photos(2)

Key Documents

AV45646

Sigma-Aldrich

Anti-NR4A3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CHN, Anti-CSMF, Anti-MINOR, Anti-NOR1, Anti-Nuclear receptor subfamily 4, group A, member 3, Anti-TEC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

68 kDa

species reactivity

mouse, rat, rabbit, pig, human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR4A3(8013)

Immunogen

Synthetic peptide directed towards the middle region of human NR4A3

Application

Anti-NR4A3 antibody produced in rabbit is suitable for western blotting at a concentration of 0.25μg/ml.

Biochem/physiol Actions

Nuclear receptor subfamily 4, group A, member 3 (NR4A3; NOR1) is an orphan receptor belonging to the steroid-thyroid hormone-retinoid receptor superfamily. It binds the NGFI-B Response Element (NBRE) and acts as transcriptional activator. NR4A3 is a regulator of mast cell function, inflammation and insulin gene expression.

Sequence

Synthetic peptide located within the following region: KCLSVGMVKEVVRTDSLKGRRGRLPSKPKSPLQQEPSQPSPPSPPICMMN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ankita Saini et al.
Scientific reports, 8(1), 2296-2296 (2018-02-06)
Mycobacterium tuberculosis instigates interactions with host factors to promote its survival within the host inimical conditions. Among such factors, nuclear receptors (NRs) seem to be promising candidates owing to their role in bacterial pathogenesis. However, only few members of NR
Weina Gao et al.
PloS one, 9(3), e91462-e91462 (2014-03-19)
NR4A3/NOR-1 is a member of the NR4A orphan nuclear receptor subfamily, which contains early response genes that sense and respond to a variety of stimuli in the cellular environment. The role of NR4A3 in insulin expression in pancreatic beta cells
Gianni Garcia-Faroldi et al.
PloS one, 9(2), e89311-e89311 (2014-03-04)
Nuclear receptor 4a3 (Nr4a3) is a transcription factor implicated in various settings such as vascular biology and inflammation. We have recently shown that mast cells dramatically upregulate Nuclear receptor 4a3 upon activation, and here we investigated the functional impact of
Takashi Nomiyama et al.
The Journal of biological chemistry, 281(44), 33467-33476 (2006-09-02)
Members of the nuclear hormone receptor superfamily function as key transcriptional regulators of inflammation and proliferation in cardiovascular diseases. In addition to the ligand-dependent peroxisome proliferator-activated receptors and liver X receptors, this family of transcription factors includes a large number

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service