Skip to Content
Merck
All Photos(3)

Key Documents

AV44249

Sigma-Aldrich

Anti-PTCH1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Patched homolog 1 (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

37 kDa

species reactivity

dog, bovine, horse, human, mouse, guinea pig, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTCH1(5727)

General description

Protein patched homolog 1 (PTCH1) is the receptor for secreted sonic hedgehog ligand and belongs to the patched gene family. It is mapped to human chromosome 9q22.32. Ptch1 has 12 transmembrane regions, intracellular middle-loop domain (MLD) and C-terminal domain (CTD) essential for oligomerization.

Immunogen

Synthetic peptide directed towards the C terminal region of human PTCH1

Application

Anti-PTCH1 antibody produced in rabbit has been used in Flow cytometry/Cell sorting.[1] It is suitable for western blotting at a concentration of 0.25μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Flow cytometry/Cell sorting (1 paper)

Biochem/physiol Actions

Mutations in protein patched homolog 1(PTCH1) is implicated in autosomal dominant disorder called the Gorlin syndrome, resulting in developmental abnormalities.
PTCH1 (patched 1; PTC) activates the sonic hedgehog (Hh) signaling pathway that is involved in the development of embryonic structures. PTCH1 is a tumor suppressor that maintains the activity of GLI1, the downstream target of Hh signaling. Mutations and methylation status in PTCH1 gene result in tumorigenesis and malignancy in cancers including colorectal, gastric, melanoma and osteosarcoma.

Sequence

Synthetic peptide located within the following region: TILGVLNGLVLLPVLLSFFGPYPEVSPANGLNRLPTPSPEPPPSVVRFAM

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jon H Chung et al.
Oncotarget, 4(12), 2208-2211 (2013-12-26)
Hedgehog (Hh) signaling is largely suppressed in the normal differentiated tissues of the adult but activated in many cancers. The Hh pathway can either be activated by the expression of Hh ligands, or by mutations that cause constitutive, ligand-independent signaling.
Swan Hwang et al.
PloS one, 8(7), e68271-e68271 (2013-08-13)
The sonic hedgehog (Shh) signaling pathway is necessary for a variety of development and differentiation during embryogenesis as well as maintenance and renascence of diverse adult tissues. However, an abnormal activation of the signaling pathway is related to various cancers.
A novel PTCH1 mutation in a patient with Gorlin syndrome
Okamoto N, et al.
Human Genome Variation, 1, 14022-14022 (2014)
Coincident PTCH and BRCA1 germline mutations in a patient with nevoid basal cell carcinoma syndrome and familial breast cancer.
Reifenberger J, et al.
The Journal of Investigative Dermatology, 116(3), 472-474 (2001)
Yun Zuo et al.
Experimental and therapeutic medicine, 6(6), 1365-1368 (2013-11-21)
The aim of this study was to investigate the correlation between patched 1 (PTCH1) expression and its methylation in a human gastric cancer cell line, in order to provide new information regarding carcinogenesis and the development of gastric cancer. Quantitative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service