Skip to Content
Merck
All Photos(1)

Key Documents

AV35645

Sigma-Aldrich

Anti-CEBPG antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CCAAT/enhancer binding protein (C/EBP), γ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

16 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CEBPG(1054)

Immunogen

Synthetic peptide directed towards the N terminal region of human CEBPG

Biochem/physiol Actions

CCAAT/enhancer binding protein (C/EBP), γ (CEBPG) is a transcription factor that regulates transcription mediated by CCAAT/enhancer element. CEBPG mediates wound repair and cell migration by affecting the EGFR-mediated signaling. It has been reported to be deregulated in acute myeloid leukemia resulting in differentiation arrest.

Sequence

Synthetic peptide located within the following region: PGVNGISVIHTQAHASGLQQVPQLVPAGPGGGGKAVAPSKQSKKSSPMDR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberta Melchionna et al.
The Journal of investigative dermatology, 132(7), 1908-1917 (2012-03-23)
We aimed at identifying novel regulators of skin wound healing (WH), in an epidermal scratch WH assay, by a small interfering RNA (siRNA) silencing approach. Several transcription factors have been previously reported to affect wound repair. We here show that
Meritxell Alberich-Jordà et al.
The Journal of clinical investigation, 122(12), 4490-4504 (2012-11-20)
C/EBPs are a family of transcription factors that regulate growth control and differentiation of various tissues. We found that C/EBPγ is highly upregulated in a subset of acute myeloid leukemia (AML) samples characterized by C/EBPα hypermethylation/silencing. Similarly, C/EBPγ was upregulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service