Skip to Content
Merck
All Photos(2)

Key Documents

AV35098

Sigma-Aldrich

Anti-KCNN4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-IK1, Anti-IKCA1, Anti-KCA4, Anti-KCa3.1, Anti-SK4, Anti-Small conductance calcium-activated channel, subfamily N, member 4, Anti-hIKCa1, Anti-hKCa4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

48 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNN4(3783)

General description

Potassium intermediate/small conductance calcium-activated channel, subfamily N, member 4, KCNN4, is a component of a potassium channel activated by calcium. It is critically involved in calcium ion transport and cell homeostasis.

Immunogen

Synthetic peptide directed towards the C terminal region of human KCNN4

Application

Anti-KCNN4 antibody produced in rabbit has been used in:
  • western blot analysis
  • immunofluorescence
  • staining
  • immunohistochemistry

Biochem/physiol Actions

KCNN4 (potassium calcium-activated channel subfamily N member 4) is part of a potentially heterotetrameric voltage-independent potassium channel that is activated by intracellular calcium. Activation is followed by membrane hyperpolarization, which promotes calcium influx. KCNN4 may be part of the predominant calcium-activated potassium channel in T-lymphocytes. Activation is followed by membrane hyperpolarization, which promotes calcium influx. The encoded protein may be part of the predominant calcium-activated potassium channel in T-lymphocytes. This gene is similar to other KCNN family potassium channel genes, but it differs enough to possibly be considered as part of a new subfamily.

Sequence

Synthetic peptide located within the following region: DLQQNLSSSHRALEKQIDTLAGKLDALTELLSTALGPRQLPEPSQQSK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The potassium channel KCa3. 1 constitutes a pharmacological target for neuroinflammation associated with ischemia/reperfusion stroke
Chen YJ, et al.
Journal of Cerebral Blood Flow and Metabolism, 36(12), 2146-2161 (2016)
Kathrin Brömmel et al.
ChemMedChem, 15(24), 2462-2469 (2020-10-13)
The Ca2+ activated potassium channel 3.1 (KCa 3.1) is involved in critical steps of the metastatic cascade, such as proliferation, migration, invasion and extravasation. Therefore, a fast and efficient protocol for imaging of KCa 3.1 channels was envisaged. The novel
D Strøbæk et al.
British journal of pharmacology, 168(2), 432-444 (2012-08-16)
The K(Ca) 3.1 channel is a potential target for therapy of immune disease. We identified a compound from a new chemical class of K(Ca) 3.1 inhibitors and assessed in vitro and in vivo inhibition of immune responses. We characterized the
Bradykinin-induced chemotaxis of human gliomas requires the activation of KCa3. 1 and ClC-3
Cuddapah VA, et al.
The Journal of Neuroscience, 33(4), 1427-1440 (2013)
Javier Lozano-Gerona et al.
PloS one, 15(3), e0222619-e0222619 (2020-03-10)
Ion channels have recently attracted attention as potential mediators of skin disease. Here, we explored the consequences of genetically encoded induction of the cell volume-regulating Ca2+-activated KCa3.1 channel (Kcnn4) for murine epidermal homeostasis. Doxycycline-treated mice harboring the KCa3.1+-transgene under the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service