Skip to Content
Merck
All Photos(1)

Key Documents

AV33704

Sigma-Aldrich

Anti-MLL4 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Myeloid/lymphoid or mixed-lineage leukemia 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

64 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MLL4(9757)

General description

Myeloid/lymphoid or mixed-lineage leukemia 4 (MLL4) is a histone-lysine N-methyltransferase widely expressed in the nucleus of multiple cell types. It contains a CXXC zinc finger, three PHD zinc fingers, two FY-rich domains, and a SET domain.

Immunogen

Synthetic peptide directed towards the N terminal region of human MLL4

Biochem/physiol Actions

MLL4 a protein which contains multiple domains including a CXXC zinc finger, three PHD zinc fingers, two FY-rich domains, and a SET (suppressor of variegation, enhancer of zeste, and trithorax) domain. The SET domain is a conserved C-terminal domain that characterizes proteins of the MLL (mixed-lineage leukemia) family. MLL4 is ubiquitously expressed in adult tissues. It is also amplified in solid tumor cell lines, and may be involved in human cancer

Sequence

Synthetic peptide located within the following region: WAGPRVQRGRGRGRGRGWGPSRGCVPEEESSDGESDEEEFQGFHSDEDVA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K T FitzGerald et al.
Genomics, 59(2), 187-192 (1999-07-20)
We have identified a gene at chromosome band 19q13.1, which is closely related to MLL. MLL is located in a region of chromosome 11q23 that has partial synteny with chromosome 19q. We have named this gene at 19q13.1, MLL2. MLL2
Khairul I Ansari et al.
Journal of molecular biology, 411(2), 334-349 (2011-06-21)
Homeobox (HOX)-containing gene HOXC6 is a critical player in mammary gland development and milk production, and is overexpressed in breast and prostate cancers. We demonstrated that HOXC6 is transcriptionally regulated by estrogen (E2). HOXC6 promoter contains two putative estrogen response
Arunoday Bhan et al.
Journal of molecular biology, 425(19), 3707-3722 (2013-02-05)
HOTAIR (HOX antisense intergenic RNA) is a long noncoding RNA (lncRNA) that is transcribed from the antisense strand of homeobox C gene locus in chromosome 12. HOTAIR coordinates with chromatin-modifying enzymes and regulates gene silencing. It is overexpressed in various
Imran Hussain et al.
Biochimica et biophysica acta, 1849(6), 697-708 (2015-03-01)
HOXC6 is a homeobox-containing gene associated with mammary gland development and is overexpressed in variety of cancers including breast and prostate cancers. Here, we have examined the expression of HOXC6 in breast cancer tissue, investigated its transcriptional regulation via estradiol
Arunoday Bhan et al.
The Journal of steroid biochemistry and molecular biology, 141, 160-170 (2014-02-19)
Antisense transcript, long non-coding RNA HOTAIR is a key player in gene silencing and breast cancer and is transcriptionally regulated by estradiol. Here, we have investigated if HOTAIR expression is misregulated by bisphenol-A (BPA) and diethylstilbestrol (DES). Our findings demonstrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service