Skip to Content
Merck
All Photos(2)

Key Documents

A9292

Sigma-Aldrich

Anti-Horse IgG (whole molecule)−Peroxidase antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Rabbit Anti-Horse IgG (whole molecule)−HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

peroxidase conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:15,000
dot blot: 1:100,000-1:200,000 (chemiluminescent)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:150

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Horse IgGs have seven subclasses ranging from IgG1 to IgG7. Equine IgG antibodies mainly regulate mucosal and systemic immunological responses and thereby, provide protection against disease-causing pathogens such as Streptococcus equi, and the horse flu virus. Horse IgG may also function to control the advancement of EHV-1 infection . Anti-horse IgG (whole molecule)−peroxidase antibody is specific for IgG in horses.

Immunogen

Horse IgG

Application

Anti-horse IgG (whole molecule)−peroxidase antibody may be used in dot blot (1:8,000-1:10,000), immunohistochemistry (formalin-fixed, paraffin-embedded sections, at 1:150 dilution) and direct ELISA (1:15,000).
The level of S2-specific antibodies in test samples of horse serum was determined by ELISA using HRP-conjugated rabbit anti-horse IgG at a 1:35000 dilution with incubation for 45 minutes at room temperature.

Biochem/physiol Actions

IgG, a monoclonal antibody can be cleaved at the hinge region by nonspecific proteases like papain and pepsin. This can result in univalent Fab (antigen-binding fragments) fragments or bivalent F(ab′)2 fragments.  These two enzymes have a broad substrate specificity resulting in heterogenous fragments.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 0.05% MIT

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sha Jin et al.
Clinical and diagnostic laboratory immunology, 11(6), 1120-1129 (2004-11-13)
We recently reported a highly protective attenuated live virus vaccine for equine infectious anemia virus (EIAV) based on a proviral construct (EIAVUKDeltaS2) with a genetically engineered mutation in the viral S2 gene that eliminates expression of this accessory protein. While
Narongsak Chaiyabutr et al.
Veterinary world, 15(10), 2365-2375 (2022-11-26)
African horse sickness (AHS) is a non-contagious, high mortality, and insect-borne disease caused by a double-stranded RNA virus from the genus Orbivirus. The study aimed to develop inactivated vaccines serotype 1 inactivated AHS vaccine (IAV) and to compare the effect
Ana Luísa Soares de Miranda et al.
Frontiers in veterinary science, 9, 852917-852917 (2022-06-18)
Loxosceles spp. (brown spiders) bites are responsible for the development of a syndrome consisting mainly of dermonecrotic lesions, and also systemic effects. Rabbits are one of the main experimental models used for better understanding the systemic and local effects of
Sabrina de Almeida Lima et al.
Frontiers in immunology, 9, 653-653 (2018-04-19)
Loxoscelism is the most important form of araneism in South America. The treatment of these accidents uses heterologous antivenoms obtained from immunization of production animals with crude loxoscelic venom. Due to the scarcity of this immunogen, new alternatives for its
Matías N Sánchez et al.
Toxicon : official journal of the International Society on Toxinology, 164, 71-81 (2019-04-19)
Most colubrid snake venoms have been poorly studied, despite the fact that they represent a great resource for biological, ecological, toxinological and pharmacological research. Herein, we explore the venom delivery system of the Aesculapian False Coral Snake Erythrolamprus aesculapii as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service