Skip to Content
Merck
All Photos(4)

Key Documents

A9013

Sigma-Aldrich

1,5-Bis(4-allyldimethylammoniumphenyl)pentan-3-one dibromide

Synonym(s):

BW 284c51

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C27H38N2OBr2
CAS Number:
Molecular Weight:
566.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

solubility

water: 19.60-20.40 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

Br.C[N](C)(CC=C)c1ccc(CCC(=O)CCc2ccc(cc2)[N](C)(C)CC=C)cc1

InChI

1S/C27H38N2O.BrH/c1-7-21-28(3,4)25-15-9-23(10-16-25)13-19-27(30)20-14-24-11-17-26(18-12-24)29(5,6)22-8-2;/h7-12,15-18H,1-2,13-14,19-22H2,3-6H3;1H

InChI key

XBXRQUYORADPMX-UHFFFAOYSA-N

Gene Information

human ... ACHE(43)

Biochem/physiol Actions

Selective acetylcholinesterase inhibitor.

Features and Benefits

This compound is featured on the Acetylcholine Synthesis and Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Olivera-Bravo et al.
British journal of pharmacology, 144(1), 88-97 (2005-01-13)
This work was aimed to determine if 1,5-bis(4-allyldimethylammoniumphenyl)pentan-3-one dibromide (BW284c51), the most selective acetylcholinesterase inhibitor (AchEI), affects the nicotinic acetylcholine (Ach) receptor (AchR) function. Purified Torpedo nicotinic AchRs were injected into Xenopus laevis oocytes and BW284c51 effects on Ach- and
Kevin B Temeyer et al.
Veterinary parasitology, 172(1-2), 114-121 (2010-05-11)
Rhipicephalus (Boophilus) microplus cDNAs, BmAChE1, BmAChE2, and BmAChE3, were previously identified as presumptively encoding acetylcholinesterases (AChEs), but biochemical identity was confirmed only for recombinant BmAChE3. In the present study, four recombinant BmAChE1 constructs and single recombinant constructs of BmAChE2 and
M Monteiro et al.
Ecotoxicology and environmental safety, 62(3), 341-347 (2005-10-12)
In recent years biomarkers have been widely used for the assessment of effects and/or exposure to environmental contaminants. One of the most frequently used biomarkers is the inhibition of cholinesterases (ChE), which is a useful indicator of organophosphate and carbamate
M Isabel Arufe et al.
Aquatic toxicology (Amsterdam, Netherlands), 84(3), 328-336 (2007-08-11)
Assessment of cholinesterase (ChE) inhibition is widely used as a specific biomarker for evaluating the exposure and effects of non-target organisms to anticholinesterase agents. Cholinesterase and carboxylesterase activities have been measured in larvae of gilthead seabream, Sparus aurata, during the
Andrea R Durrant et al.
Frontiers in molecular neuroscience, 5, 73-73 (2012-06-23)
The cholinesterases, acetylcholinesterase (AChE), and butyrylcholinesterase (BChE) (pseudocholinesterase), are abundant in the nervous system and in other tissues. The role of AChE in terminating transmitter action in the peripheral and central nervous system is well understood. However, both knowledge of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service