Skip to Content
Merck
All Photos(1)

Key Documents

A8332

Sigma-Aldrich

p-Aminobenzamidine–Agarose

(1:1 suspension in a 1.0 M NaCl + 20% ethanol solution)

Synonym(s):

pABA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106514
NACRES:
NA.56

biological source

plant

Quality Level

form

(1:1 suspension in a 1.0 M NaCl + 20% ethanol solution)

technique(s)

protein purification: suitable

matrix

cross-linked 6% beaded agarose

matrix activation

epichlorohydrin

matrix attachment

amino to carboxyl of spacer

matrix spacer

9 atoms

capacity

≥5 mg/mL binding capacity (trypsin)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

p-Aminobenzamidine Agarose (pABA) can be used as an affinity ligand to bind and remove serine proteases in addition to Ca2+-binding proteins. It has also been used in medical applications as a tool for the purification of non-protease proteins.

Physical form

1:1 suspension in a 1.0 M NaCl + 20% ethanol solution

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gulay Bayramoglu et al.
Bioprocess and biosystems engineering, 35(3), 423-431 (2011-08-20)
In this work, a new methodology is developed for selection of affinity ligands towards the enzyme "trypsin" using quartz crystals microbalance (QCM) technique. To achieve this goal, the surface amination of gold plated QCM crystals was achieved in 13.56 MHz
Massimo D'Agostino et al.
Nature, 551(7682), 634-638 (2017-11-02)
Membrane fusion in eukaryotic cells mediates the biogenesis of organelles, vesicular traffic between them, and exo- and endocytosis of important signalling molecules, such as hormones and neurotransmitters. Distinct tasks in intracellular membrane fusion have been assigned to conserved protein systems.
Paola Fortugno et al.
Journal of cell science, 115(Pt 3), 575-585 (2002-02-28)
Survivin is a member of the inhibitor of apoptosis gene family that has been implicated in both apoptosis inhibition and regulation of mitosis. However, the subcellular distribution of survivin has been controversial and variously described as a microtubule-associated protein or
Ya-Chi Su et al.
The Journal of biological chemistry, 286(36), 31904-31914 (2011-07-23)
To select residues in coagulation factor XIa (FXIa) potentially important for substrate and inhibitor interactions, we examined the crystal structure of the complex between the catalytic domain of FXIa and the Kunitz protease inhibitor (KPI) domain of a physiologically relevant
Tomas Erban
PloS one, 6(8), e22860-e22860 (2011-08-31)
p-aminobenzamidine (p-ABA) is used as a ligand in the purification of many serine proteases and in their removal from heterogeneous samples. Moreover, p-ABA has a potent ability to bind Ca(2+)-binding proteins. The binding ability and use of p-ABA in purification

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service