Skip to Content
Merck
All Photos(2)

Key Documents

A8138

Sigma-Aldrich

8-(6-Aminohexyl)aminoadenosine 3′:5′-cyclic monophosphate

~98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26N7O6P
CAS Number:
Molecular Weight:
443.39
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

~98%

Quality Level

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

NCCCCCCNc1nc2c(N)ncnc2n1[C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O

InChI

1S/C16H26N7O6P/c17-5-3-1-2-4-6-19-16-22-10-13(18)20-8-21-14(10)23(16)15-11(24)12-9(28-15)7-27-30(25,26)29-12/h8-9,11-12,15,24H,1-7,17H2,(H,19,22)(H,25,26)(H2,18,20,21)/t9-,11-,12-,15-/m1/s1

InChI key

WCWLOZYNRVLFOG-SDBHATRESA-N

Related Categories

Application

Cyclic AMP analog which shows partially selective agonist activity with Type I cAMP-dependent protein kinase isozyme.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anirban Chakraborty et al.
Methods in enzymology, 472, 19-30 (2010-06-29)
We describe the synthesis of phosphine derivatives of three fluorescent probes that have a brightness and photostability suitable for single-molecule fluorescence spectroscopy and microscopy: Alexa488, Cy3B, and Alexa647. In addition, we describe procedures for use of these reagents in azide-specific
Jong Seol Yuk et al.
Biosensors & bioelectronics, 26(7), 3213-3218 (2011-01-25)
We present a novel approach to the enhancement of surface plasmon-coupled emission (SPCE) using surface plasmon excitation in a bimetal (Ag/Au) layer and we validate the enhancement by presenting the results of a model human IgG immunoassay. Theoretical calculations using
Jody L Swift et al.
Biointerphases, 5(4), 139-148 (2011-01-12)
Fluorescence microscopy methods including total internal reflection fluorescence and confocal laser scanning microscopy have played a major role in modern cell biology research by permitting imaging of fluorescently tagged macromolecules in living cells. These methods are often used to examine
Hamed Shadpour et al.
ACS applied materials & interfaces, 2(4), 1086-1093 (2010-04-29)
A method to perform in situ roughening of arrays of microstructures weakly adherent to an underlying substrate was presented. SU8, 1002F, and polydimethylsiloxane (PDMS) microstructures were roughened by polishing with a particle slurry. The roughness and the percentage of dislodged
Daniel C Propheter et al.
Molecular bioSystems, 7(7), 2114-2117 (2011-05-21)
Herein we describe the orientation of GST-tagged lectins on NHS-activated slides via a one-step deposition of the protein and a glutathione (GSH) scaffold. This technology overcomes the need for a premade GSH-surface to orient GST-tagged proteins, enabling us to rapidly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service