Skip to Content
Merck
All Photos(3)

Key Documents

A5880

Sigma-Aldrich

D-Amphetamine hemisulfate salt

Synonym(s):

(+)-α-Methylphenethylamine hemisulfate salt, Dextroamphetamine hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N · 0.5 H2SO4
CAS Number:
Molecular Weight:
184.25
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

solubility

H2O: soluble
ethanol: soluble

application(s)

forensics and toxicology

SMILES string

OS(O)(=O)=O.CC(N)Cc1ccccc1.CC(N)Cc2ccccc2

InChI

1S/2C9H13N.H2O4S/c2*1-8(10)7-9-5-3-2-4-6-9;1-5(2,3)4/h2*2-6,8H,7,10H2,1H3;(H2,1,2,3,4)/t2*8-;/m00./s1

InChI key

PYHRZPFZZDCOPH-QXGOIDDHSA-N

Gene Information

human ... SLC18A2(6571)

Looking for similar products? Visit Product Comparison Guide

Application

D-Amphetamine hemisulfate salt has been used in oxidative fermentative (OF) test, and exploratory test.

Biochem/physiol Actions

D-Amphetamine is an active isomer of amphetamine. It induces release of catecholamines and serotonin by displacing the monoamines from their vesicular storage sites, which blocks catecholamine reuptake.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy J O'Neal et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 45(8), 1251-1262 (2019-11-21)
Opioid addiction has been declared a public health emergency, with fatal overdoses following relapse reaching epidemic proportions and disease-associated costs continuing to escalate. Relapse is often triggered by re-exposure to drug-associated cues, and though the neural substrates responsible for relapse
A M Barr et al.
Pharmacology, biochemistry, and behavior, 64(3), 597-604 (1999-11-05)
The present study sought to determine the effect of withdrawal from an escalating dose schedule of d-amphetamine on sexual behavior in male rats. Tests were conducted every 5 days until stable levels of sexual behavior were obtained. With repeated testing
Cedric Jackson et al.
Behavioural brain research, 361, 131-138 (2018-12-15)
Although scientific research using mammalian models has made great strides in uncovering the enigmatic neural and molecular mechanisms orchestrating the state of drug addiction, a complete understanding has thus far eluded researchers. The complexity of the task has led to
J Cartmell et al.
The Journal of pharmacology and experimental therapeutics, 291(1), 161-170 (1999-09-22)
Previous animal studies have indicated that drugs targeted at metabotropic glutamate (mGlu) receptors may be useful for treatment of psychosis. In this article, the effects of the novel, potent, and selective mGlu2/3 receptor agonists LY354740 and LY379268, and the clinically
Stephanie A Carmack et al.
Neuroscience letters, 468(1), 18-22 (2009-10-27)
Much research is focused on developing novel drugs to improve memory. In particular, psychostimulants have been shown to enhance memory and have a long history of safe use in humans. In prior work, we have shown that very low doses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service