Skip to Content
Merck
All Photos(2)

Key Documents

A5638

Sigma-Aldrich

Adenosine 2′,5′-diphosphate sodium salt

≥95%

Synonym(s):

2′-Phosphoadenosine 5′-phosphate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N5O10P2 · xNa+
CAS Number:
Molecular Weight:
427.20 (free acid basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Quality Level

Assay

≥95%

form

powder

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]3OP(O)(O)=O

InChI

1S/C10H15N5O10P2.Na/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(25-27(20,21)22)6(16)4(24-10)1-23-26(17,18)19;/h2-4,6-7,10,16H,1H2,(H2,11,12,13)(H2,17,18,19)(H2,20,21,22);

InChI key

OHPUPSOOFQPMRP-UHFFFAOYSA-N

Related Categories

Application

Adenosine 2′,5′-diphosphate may be used to study the mechanisms and activities of these components. Adenosine 2′,5′-diphosphate may be used to prepare adenosine 2′,5′-diphosphate agarose for affinity chromatography purification of enzymes such as NADPH-cytochrome P450 reductase and glutathione reductase.

Biochem/physiol Actions

Adenosine 2′,5′-diphosphate (A2P5P) is a competitive antagonist of the P2Y1 receptor and a non-selective antagonist of the platelet P2X1 ion channel. A2P5P prevents ADP-induced platelet aggregation and deformation but it does not affect ADP-induced inhibition of adenylyl cyclase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R G Stevens et al.
Planta, 211(4), 537-545 (2000-10-13)
Expression in transgenic tobacco (Nicotiana tabacum L.) of a pea (Pisum sativum L.) GOR2 cDNA, encoding an isoform of glutathione reductase (GOR2), resulted in a 3- to 7-fold elevation of total foliar glutathione reductase (GR) activity. The enzyme encoded by
F Moccia et al.
Microvascular research, 61(3), 240-252 (2001-05-05)
Intracellular Ca2+ signals elicited by nucleotide agonists were investigated in primary cultures of rat cardiac microvascular endothelial cells using the fura-2 technique. UTP increased the intracellular [Ca2+] in 94% of the cells, whereas 2MeSATP was active in 32%. The rank
H Schwarz et al.
Plant biology (Stuttgart, Germany), 11(3), 300-306 (2009-05-28)
Solubilised NADPH-cytochrome P450 reductase (CPR) was purified from the microsomal fraction of centaury (Centaurium erythraea) cell cultures by Q-anion exchange chromatography and affinity chromatography on adenosine 2',5'-diphosphate agarose. SDS-PAGE demonstrated the presence of three CPR isoforms with molecular masses of
J L Boyer et al.
Molecular pharmacology, 50(5), 1323-1329 (1996-11-01)
Although P2 receptors mediate a myriad of physiological effects of extracellular adenine nucleotides, study of this broad class of receptors has been compromised by a lack of P2 receptor-selective antagonist molecules. The adenine nucleotide-promoted inositol lipid hydrolysis response of turkey
The platelet P2 receptors
Platelets, 259-277 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service