Skip to Content
Merck
All Photos(1)

Key Documents

A5356

Sigma-Aldrich

N-Me-Abz-Amyloid β/A4 Protein Precursor770 (708-715)-Lys(Dnp)-D-Arg-D-Arg-D-Arg amide trifluoroacetate salt

≥96% (HPLC), lyophilized powder

Synonym(s):

N-Me-Abz-Amyloid β-Protein (37-44)-Lys(Dnp)-D-Arg-D-Arg-D-Arg amide trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H116N26O18 · xC2HF3O2
Molecular Weight:
1609.83 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.32

Pricing and availability is not currently available.

Quality Level

Assay

≥96% (HPLC)

form

lyophilized powder

composition

Peptide content, 92% HPLC

solubility

H2O: >1 mg/mL

shipped in

dry ice

storage temp.

−20°C

Amino Acid Sequence

N-Me-Abz-Gly-Gly-Val-Val-Ile-Ala-Thr-Val-Lys(Dnp)-D-Arg-D-Arg-D-Arg-NH2

Biochem/physiol Actions

A novel intramolecularly quenched fluorescent, presenilin-dependent substrate for assaying γ -secretase activity. It has been used for partial purification and characterization of γ-secretase from post-mortem human brain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Mehmet Fatih Bolukbasi et al.
    Molecular therapy. Nucleic acids, 1, e10-e10 (2013-01-25)
    Despite intensive studies, the molecular mechanisms by which the genetic materials are uploaded into microvesicles (MVs) are still unknown. This is the first study describing a zipcode-like 25 nucleotide (nt) sequence in the 3'-untranslated region (3'UTR) of mRNAs, with variants
    Okay Saydam et al.
    Molecular and cellular biology, 29(21), 5923-5940 (2009-08-26)
    Meningiomas, one of the most common human brain tumors, are derived from arachnoidal cells associated with brain meninges, are usually benign, and are frequently associated with neurofibromatosis type 2. Here, we define a typical human meningioma microRNA (miRNA) profile and

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service