Skip to Content
Merck
All Photos(1)

Key Documents

A4933

Sigma-Aldrich

Astressin

≥90% (HPLC)

Synonym(s):

[D-Phe12, Nle21,38, Glu30, Lys33]-Corticotropin Releasing Factor fragment 12-41

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C161H269N49O42
CAS Number:
Molecular Weight:
3563.16
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

Quality Level

Assay

≥90% (HPLC)

form

powder

storage temp.

−20°C

InChI

1S/C161H269N49O42/c1-23-27-41-96(189-145(239)107(52-59-123(217)218)199-152(246)114(71-84(13)14)207-157(251)126(85(15)16)208-147(241)108(53-60-124(219)220)197-140(234)102(47-38-66-179-161(172)173)193-151(245)112(69-82(9)10)204-153(247)113(70-83(11)12)205-155(249)116(74-94-77-175-79-181-94)201-134(228)95(163)72-92-39-30-29-31-40-92)135(229)182-88(19)130(224)186-100(45-36-64-177-159(168)169)136(230)183-89(20)131(225)188-106(51-58-122(215)216)144(238)195-104(49-56-119(165)212)146(240)202-110(67-80(5)6)149(243)185-90(21)132(226)187-103(48-55-118(164)211)143(237)196-105-50-57-121(214)176-63-35-33-44-99(192-154(248)115(73-93-76-174-78-180-93)200-133(227)91(22)184-137(105)231)142(236)206-117(75-120(166)213)156(250)194-101(46-37-65-178-160(170)171)139(233)190-98(43-32-34-62-162)141(235)203-111(68-81(7)8)150(244)191-97(42-28-24-2)138(232)198-109(54-61-125(221)222)148(242)210-128(87(18)26-4)158(252)209-127(129(167)223)86(17)25-3/h29-31,39-40,76-91,95-117,126-128H,23-28,32-38,41-75,162-163H2,1-22H3,(H2,164,211)(H2,165,212)(H2,166,213)(H2,167,223)(H,174,180)(H,175,181)(H,176,214)(H,182,229)(H,183,230)(H,184,231)(H,185,243)(H,186,224)(H,187,226)(H,188,225)(H,189,239)(H,190,233)(H,191,244)(H,192,248)(H,193,245)(H,194,250)(H,195,238)(H,196,237)(H,197,234)(H,198,232)(H,199,246)(H,200,227)(H,201,228)(H,202,240)(H,203,235)(H,204,247)(H,205,249)(H,206,236)(H,207,251)(H,208,241)(H,209,252)(H,210,242)(H,215,216)(H,217,218)(H,219,220)(H,221,222)(H4,168,169,177)(H4,170,171,178)(H4,172,173,179)/t86-,87-,88-,89-,90-,91-,95+,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,112-,113-,114-,115-,116-,117-,126-,127-,128-/m0/s1

InChI key

HPYIIXJJVYSMCV-MGDXKYBTSA-N

Gene Information

human ... CRH(1392)
mouse ... CRH(12918)
rat ... CRH(81648)

Amino Acid Sequence

Phe-His-Leu-Leu-Arg-Glu-Val-Leu-Glu-Nle-Ala-Arg-Ala-Glu-Gln-Leu-Ala-Gln-cyclo-(Glu-Ala-His-Lys)-Asn-Arg-Lys-Leu-Nle-Glu-Ile-Ile-NH2

Application

Astressin has been used as a CRF-receptor antagonist to study its effects on the decrease in the Schaffer collateral-CA1 field potential induced by hypoxia/hypoglycemia (ischemia) in rat hippocampal slices.

Biochem/physiol Actions

Astressin is a non-selective corticotropin releasing factor (CRF)-receptor antagonist. It potentially blocks CRF-induced adrenocorticotropic hormone (ACTH) release. Astressin effectively deceases delayed effects of chronic social isolation such as anxiety and fobial states in rats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Behavioral effects of central administration of the novel CRF antagonist astressin in rats
Spina MG, et al.
Neuropsychopharmacology, 22(3), 230-239 (2000)
P D Shabanov et al.
Eksperimental'naia i klinicheskaia farmakologiia, 74(7), 3-8 (2011-09-08)
Bipolar electrodes were implanted in the lateral hypothalamus in a group of 44 Wistar male rats in order to study self-stimulation reaction in the Skinner box. Simultaneously, microcanules were implanted into the central nucleus of the amygdala to inject the
Young Ju Cho et al.
Journal of Korean medical science, 26(2), 279-283 (2011-02-03)
Corticotrophin-releasing factor (CRF) plays a major role in coordinating stress responses. We aimed to test whether blocking endogenous CRF activity can prevent the stress-induced dilation of intercellular spaces in esophageal mucosa. Eighteen adult male rats were divided into 3 groups:
L Yang et al.
Physiology & behavior, 104(5), 886-892 (2011-06-10)
Rapid eye movement sleep (REM) is increased after controllable stress (modeled by escapable footshock, ES) and decreased after uncontrollable stress (modeled by inescapable footshock, IS). Decreases in REM after IS are exacerbated by corticotropin releasing factor (CRF) and attenuated by
Kabirullah Lutfy et al.
Journal of neurochemistry, 120(6), 1108-1116 (2011-12-24)
Corticosterone-releasing hormone (CRH) and arginine vasopressin (AVP) are crucial components of the hypothalamic-pituitary-adrenal axis that stimulates the release of adrenocorticotropic hormone from the pituitary and mediate the stress response. CRH binds to two subtypes of CRH receptors (CRH-R1 and CRH-R2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service