Skip to Content
Merck
All Photos(3)

Key Documents

A4626

Sigma-Aldrich

1-Acetyl-2-phenylhydrazine

≥98%

Synonym(s):

N-Acetyl-N′-phenylhydrazine, acetylphenylhydrazine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHNHC6H5
CAS Number:
Molecular Weight:
150.18
EC Number:
MDL number:
UNSPSC Code:
12352127
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98%

form

(Powder with Chunk(s) or Crystals with Chunk(s))

mp

128-131 °C (lit.)

SMILES string

CC(=O)NNc1ccccc1

InChI

1S/C8H10N2O/c1-7(11)9-10-8-5-3-2-4-6-8/h2-6,10H,1H3,(H,9,11)

InChI key

UICBCXONCUFSOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Acetyl-2-phenylhydrazine (AcPhHZ) which reacts with oxyhaemoglobin to form free radicals may be used to induce haemolytic anaemia for studies of the hematopoietic system.

Biochem/physiol Actions

1-Acetyl-2-phenylhydrazine (AcPhHZ) is a vascular tumor initiator used in experimental animal model.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H Ohno et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 32(2), 269-277 (1996-08-01)
A previous study revealed that DQ-2511, a new gastroprokinetic drug, induced hemolytic anemia together with increased Heinz body formation, preceded by a marked decrease in erythrocyte reduced glutathione (GSH) content, after 2 weeks of dosing onward in dogs. In this
G Bartosz et al.
Clinica chimica acta; international journal of clinical chemistry, 198(3), 239-243 (1991-05-15)
Erythrocytes of Down syndrome patients, having increased activity of superoxide dismutase and glutathione peroxidase, showed a similar increase in intracellular proteolysis upon stimulation with 1 mmol/l phenylhydrazine than normal erythrocytes. There were no differences in the magnitude of Heinz body
Y Manabe et al.
The Journal of toxicological sciences, 25(3), 161-165 (2000-09-15)
In this study, a new simple method to measure erythrocyte fragility with stirring of diluted blood (stirring method) was introduced and evaluated with anemic rats given beta-acetylphenylhydrazine (APHZ) or clofibrate. APHZ at a dose of 40 mg/kg caused significant decreases
Simonetta Croci et al.
Anticancer research, 22(5), 2903-2906 (2003-01-18)
The end result of our research would be the development of a new diagnostic technique that could be used as a screening test, starting from a simple blood collection, a common procedure without any risk to the subject. Red blood
T Umenai et al.
The Tohoku journal of experimental medicine, 186(3), 181-191 (1999-05-29)
An increase in the number of erythroblasts can be seen to some extent in the bone marrow of rats in the early stage of experimentally induced hemolytic anemia prior to any elevation in the plasma erythropoietin (Epo) level. This observation

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service