Skip to Content
Merck
All Photos(9)

Key Documents

A3377

Sigma-Aldrich

Adenosine 5′-triphosphate disodium salt hydrate

Grade II, ≥97% (HPLC), crystalline, from microbial

Synonym(s):

ATP disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N5Na2O13P3 · xH2O
CAS Number:
Molecular Weight:
551.14 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.51

biological source

microbial

Quality Level

type

Grade II

Assay

≥97% (HPLC)

form

crystalline

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O

InChI

1S/C10H16N5O13P3.2Na.H2O/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;;1H2/q;2*+1;/p-2/t4-,6-,7-,10-;;;/m1.../s1

InChI key

NTBQNWBHIXNPRU-MSQVLRTGSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Adenosine 5′-triphosphate (ATP) is a central component of energy storage and metabolism in vivo. ATP is use in many cellular processes, respiration, biosynthetic reactions, motility, and cell division. ATP is a substrate of many kinases involved in cell signaling and of adenylate cyclase(s) that produce the second messenger cAMP. ATP provides the metabolic energy to drive metabolic pumps. ATP serves as a coenzyme in a wide array of enzymatic reactions.

Biochem/physiol Actions

P2 purinergic agonist; increases activity of Ca2+-activated K+ channels; substrate for ATP-dependent enzyme systems
P2 purinocepter agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peifa Yu et al.
Virology, 546, 109-121 (2020-05-27)
The inflammasome machinery has recently been recognized as an emerging pillar of innate immunity. However, little is known regarding the interaction between the classical interferon (IFN) response and inflammasome activation in response to norovirus infection. We found that murine norovirus
Iris R M Tébéka et al.
Langmuir : the ACS journal of surfaces and colloids, 25(3), 1582-1587 (2009-01-28)
Cellulase is an enzymatic complex which synergically promotes the degradation of cellulose to glucose. The adsorption behavior of cellulase from Trichoderma reesei onto Si wafers or amino-terminated surfaces was investigated by means of ellipsometry and atomic force microscopy (AFM) as
Zachary M March et al.
eLife, 9 (2020-12-16)
The AAA+ protein disaggregase, Hsp104, increases fitness under stress by reversing stress-induced protein aggregation. Natural Hsp104 variants might exist with enhanced, selective activity against neurodegenerative disease substrates. However, natural Hsp104 variation remains largely unexplored. Here, we screened a cross-kingdom collection
Fabian Sesterhenn et al.
PLoS biology, 17(2), e3000164-e3000164 (2019-02-23)
Throughout the last several decades, vaccination has been key to prevent and eradicate infectious diseases. However, many pathogens (e.g., respiratory syncytial virus [RSV], influenza, dengue, and others) have resisted vaccine development efforts, largely because of the failure to induce potent
Christopher J Groten et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(6), 2747-2765 (2015-02-13)
It is unknown whether neurons can dynamically control the capacity for secretion by promptly changing the number of plasma membrane voltage-gated Ca(2+) channels. To address this, we studied peptide release from the bag cell neurons of Aplysia californica, which initiate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service