Skip to Content
Merck
All Photos(1)

Key Documents

A2846

Sigma-Aldrich

A-134974 dihydrochloride hydrate

≥98% (HPLC), solid

Synonym(s):

N7-[(1′R,2′S,3′R,4′S)-2′,3′-dihydroxy-4′-aminocyclopentyl]-4-amino-5-iodopyrrolopyrimidine dihydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14IN5O2 · 2HCl · xH2O
CAS Number:
Molecular Weight:
448.09 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

off-white to light tan

solubility

H2O: soluble

originator

Abbott

SMILES string

O.Cl.Cl.N[C@H]1C[C@H]([C@H](O)[C@@H]1O)n2cc(I)c3c(N)ncnc23

InChI

1S/C11H14IN5O2.2ClH.H2O/c12-4-2-17(6-1-5(13)8(18)9(6)19)11-7(4)10(14)15-3-16-11;;;/h2-3,5-6,8-9,18-19H,1,13H2,(H2,14,15,16);2*1H;1H2/t5-,6+,8+,9-;;;/m0.../s1

InChI key

INMLNTBMPNIKQK-HWZUHWJPSA-N

Biochem/physiol Actions

A-134974 is a novel and selective adenosine kinase (AK) inhibitor with IC50 = 60 pM. Systemic A-134974 (i.p.) dose dependently reduced hyperalgesia (ED50= 1 μmol/kg) and at higher doses, reduced locomotor activity (ED50 = 16 μmol/kg). Administration of A-134974 intrathecally (i.t.) was more potent (ED50= 6 nmol) at producing antihyperalgesia than delivering the compound by intracerebralventricular (ED50 = 100 nmol, i.c.v.) or intraplantar (ED50 >300 nmol) routes. In contrast, i.c.v. administration of A-134974 was more effective in reducing locomotor activity than i.t. administration (ED50 values were 1 and >100 nmol, respectively). Increasing the pretreatment time for i.t.-delivered A-134974 caused a greater reduction in locomotor activity (ED50= 10 nmol). This was due to diffusion of A-134974 (i.t.) to supraspinal sites. These data demonstrate that the novel AK inhibitor A-134974 potently reduces thermal hyperalgesia primarily through interactions with spinal sites, whereas its ability to depress locomotor activity is predominantly mediated by supraspinal sites.
A-134974 is a novel and selective adenosine kinase inhibitor. In a rat model of neuropathic pain, intraperitoneal A-134974 reduced tactile allodynia and hyperalgesia in a dose-dependent manner (IC50 = 60 pM).

Features and Benefits

This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold under license from Abbott Laboratories.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to adenosine receptors for your research needs.

We offers many products related to adenosine receptors for your research needs.

We offers many products related to adenosine receptors for your research needs.

We offers many products related to adenosine receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service