Skip to Content
Merck
All Photos(2)

Key Documents

A254

Sigma-Aldrich

AIDA

solid

Synonym(s):

1-Amino-2,3-dihydro-1H-indene-1,5-dicarboxylic acid, 1-Aminoindan-1, 5-dicarboxylic acid, UPF 523

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO4
CAS Number:
Molecular Weight:
221.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

form

solid

Quality Level

color

white

solubility

DMSO: 2 mg/mL

SMILES string

NC1(CCc2cc(ccc12)C(O)=O)C(O)=O

InChI

1S/C11H11NO4/c12-11(10(15)16)4-3-6-5-7(9(13)14)1-2-8(6)11/h1-2,5H,3-4,12H2,(H,13,14)(H,15,16)

InChI key

LSTPKMWNRWCNLS-UHFFFAOYSA-N

Gene Information

human ... GRM1(2911)
rat ... Grm1(24414)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Selective mGluR1 metabotropic glutamate receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Joseph et al.
Molecular neurodegeneration, 12(1), 88-88 (2017-12-06)
The microtubule-associated protein Tau plays a role in neurodegeneration as well as neurogenesis. Previous work has shown that the expression of the pro-aggregant mutant Tau repeat domain causes strong aggregation and pronounced neuronal loss in the hippocampus whereas the anti-aggregant
F Moroni et al.
The Journal of pharmacology and experimental therapeutics, 281(2), 721-729 (1997-05-01)
We examined the pharmacological profile of 1-aminoindan-1,5-dicarboxylic acid (AIDA), a rigid (carboxyphenyl)glycine derivative acting on metabotropic glutamate receptors (mGluRs). In cells transfected with mGluR1a, AIDA competitively antagonized the stimulatory responses of glutamate and (1S,3R)-1-aminocyclopentane-1,3-dicarboxylic acid [(1S,3R)-ACPD] on phosphoinositide hydrolysis (pA2
Andreas Schuster et al.
Journal of clinical medicine, 9(1) (2020-01-17)
Background: Right ventricular (RV) function is a known predictor of adverse events in heart failure and following acute myocardial infarction (AMI). While right atrial (RA) involvement is well characterized in pulmonary arterial hypertension, its relative contributions to adverse events following
Friedrich-Christian Deuschle et al.
Theranostics, 10(5), 2172-2187 (2020-02-25)
Enhanced amino acid supply and dysregulated integrin signaling constitute two hallmarks of cancer and are pivotal for metastatic transformation of cells. In line with its function at the crossroads of both processes, overexpression of CD98hc is clinically observed in various
Christine E King et al.
Journal of neuroengineering and rehabilitation, 10, 77-77 (2013-07-23)
Spinal cord injury (SCI) can leave the affected individuals with paraparesis or paraplegia, thus rendering them unable to ambulate. Since there are currently no restorative treatments for this population, novel approaches such as brain-controlled prostheses have been sought. Our recent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service