Skip to Content
Merck
All Photos(1)

Key Documents

A2348

Sigma-Aldrich

Avidin−Sulforhodamine 101

lyophilized powder

Synonym(s):

Avidin-Texas Red® conjugate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

conjugate

sulforhodamine 101 conjugate

Quality Level

form

lyophilized powder

mol wt

~66,000

storage temp.

−20°C

General description

Highly cationic glycoprotein that can be used to identify mast cells.
Highly cationic glycoprotein that can selectively bind to a component in human and rodent mast cell granules in fixed-cell preparations, and can be used to identify mast cells in normal and diseased human tissue without requiring a biotinylated probe.

Application

Avidin-Sulforhodamine 101 (Avidin-Texas Red®) has been used for studying the printing of organic nanomaterials using electrospray ionization and Coulomb-force-directed assembly techniques[1]. The product has also been used for microcontact printing studies[2].
Avidin-conjugated Texas red was used to visualize biotinylated centromeric DNA in a FISHI assay.

Legal Information

Texas Red is a registered trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Printing of organic and inorganic nanomaterials using electrospray ionization and Coulomb-force-directed assembly.
Welle, A. M., and Jacobs, H. O.
Applied Physics Letters, 87(26), 263119-263119 (2005)
Protein In-Mold Patterning
Biancardo, S. B., et al.
Advanced Materials, 20(10), 1825-1829 (2008)
Leiyun Yang et al.
The New phytologist, 226(2), 507-522 (2019-12-20)
Plant immune responses need to be tightly controlled for growth-defense balance. The mechanism underlying this tight control is not fully understood. Here we identify epigenetic regulation of nucleotide-binding leucine rich repeat or Nod-Like Receptor (NLR) genes as an important mechanism

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service