Skip to Content
Merck
All Photos(1)

Key Documents

A231

Sigma-Aldrich

Arachidonyl trifluoromethyl ketone

oil, ≥97% (NMR)

Synonym(s):

1,1,1-Trifluoromethyl-6,9,12,15-heieicosatetraen-2-one, AACOCF3, Arach-CF3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31F3O
CAS Number:
Molecular Weight:
356.47
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (NMR)

form

oil

storage condition

desiccated
protect from light
under inert gas

color

yellow

solubility

DMSO: >5 mg/mL

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)C(F)(F)F

InChI

1S/C21H31F3O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(25)21(22,23)24/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-19H2,1H3/b7-6-,10-9-,13-12-,16-15-

InChI key

PLWROONZUDKYKG-DOFZRALJSA-N

Gene Information

human ... PLA2G1B(5319)

Application

Arachidonyl trifluoromethyl ketone (AACOCF3) has been used as a anandamide degradation inhibitor to study its effects in primary astrocyte cultures.

Biochem/physiol Actions

Arachidonyl trifluoromethyl ketone (AACOCF3) plays a role in the inhibition of 12-hydroxyeicosatetraenoic acid (12-HETE) and thromboxane B2 produced by the platelets.
Inhibits anandamide hydrolysis in vitro; inhibits phospholipase A2.

Features and Benefits

This compound is featured on the Phospholipase A2 page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Hygroscopic, photosensitive, air sensitive

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam C Vana et al.
Experimental neurology, 231(1), 45-55 (2011-06-21)
Inhibition of phospholipase A(2) (PLA(2)) has recently been found to attenuate the pathogenesis of experimental autoimmune encephalomyelitis (EAE), a commonly used animal model of multiple sclerosis (MS). However, the protective mechanisms that underlie PLA(2) inhibition are still not well understood.
Backil Sung et al.
Pain, 131(1-2), 121-131 (2007-02-03)
Spinal glutamate transporters (GT) have been implicated in the mechanisms of neuropathic pain; however, how spinal GT uptake activity is regulated remains unclear. Here we show that alteration of spinal arachidonic acid (AA) turnover after peripheral nerve injury regulated regional
B Koutek et al.
The Journal of biological chemistry, 269(37), 22937-22940 (1994-09-16)
Arachidonoyl ethanolamide (anandamide) is a naturally occurring brain constituent that binds to a specific brain cannabinoid receptor (CBR1). An amidase activity (anandamide amidase) in membrane fractions of brain and in cultured neuroblastoma cells rapidly degrades anandamide to arachidonic acid (Deutsch
Wen Li et al.
Biochemistry, 47(52), 13997-14008 (2008-12-10)
TCDD (dioxin) induces a rapid inflammatory response from 3T3-L1 adipocytes as judged by prominent induction of the mRNA expression of prostaglandin-endperoxide synthase 2 (Cox-2) along with other inflammation markers within 1 h. This action of TCDD is clearly antagonized by
The endogenous cannabinoid anandamide potentiates interleukin-6 production by astrocytes infected with Theiler's murine encephalomyelitis virus by a receptor-mediated pathway
Molina H, et al.
Febs Letters, 433(1-2), 139-142 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service