Skip to Content
Merck
All Photos(1)

Key Documents

A0412

Sigma-Aldrich

Anti-Mouse Polyvalent Immunoglobulins (G,A,M)−Peroxidase antibody produced in goat

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

technique(s)

direct ELISA: 1:10,000 using IgG, IgA, IgM

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Immunoglobulins are proteins produced by B cells in response to antigen and regulate response to bacteria, and viruses. IgG is known to regulate complement fixation and placental transport. IgA has a crucial role in mucosal immunity as it restricts pathogens from entering the mucosal membrane. IgM is the largest antibody having a pentameric structure which modulates polyreactivity and removes apoptotic cells,,. Goat anti-mouse polyvalent immunoglobulins (G,A,M)-peroxidase antibody binds to mouse IgG, IgA and IgM.

Application

The detection of antibodies in sera of T. gondii-infected mice was done by ELISA using peroxidase-conjugated anti-mouse polyvalent immunoglobulin antibody as the secondary. Relative binding affinities for specific Mabs were determined by an inhibition ELISA using peroxidase-conjugated goat anti-mouse polyvalent antibody diluted 1:10,000 in PBST and incubated for 1 hour at 21-23°C.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4 containing 1% bovine serum albumin with preservative.

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole Forster et al.
Journal of immunology (Baltimore, Md. : 1950), 178(11), 6941-6948 (2007-05-22)
Conditional knock-in mice expressing a histone acetyltransferase-deficient version of the transcriptional coregulator p300 exclusively in B lymphocytes die prematurely with full penetrance. The mice develop an autoimmune disease similar to systemic lupus erythematosus in its pathological manifestations, such as splenomegaly
M Chen et al.
Infection and immunity, 68(9), 4893-4899 (2000-08-19)
Formation of anti-Toxoplasma gondii HSP70 (TgHSP70) antibody cross-reactive to mouse HSP70 (mHSP70) was observed in the sera of BALB/c (a resistant strain) and C57BL/6 (B6; a susceptible strain) mice after peroral infection with T. gondii cysts of the Fukaya strain.
Hong Xin et al.
Infection and immunity, 74(7), 4310-4321 (2006-06-23)
We previously reported the enhanced resistance of monoclonal antibodies B6.1 (an immunoglobulin M [IgM]) and C3.1 (an IgG3) against experimental candidiasis. Both MAbs recognize the same fungal epitope. We have since found that a highly passaged B6.1 hybridoma (hp-B6.1) resulted
The Effect of the Herbicide Mecoprop on Heliscus lugdunensis and Its Influence on the Preferential Feeding of Gammarus Pseudolimnaeus.
S Bermingham et al.
Microbial ecology, 35(2), 199-204 (1998-12-16)
Valentina Agnolon et al.
Frontiers in immunology, 11, 586595-586595 (2020-12-01)
The humoral responses of Ebola virus (EBOV) survivors mainly target the surface glycoprotein GP, and anti-GP neutralizing antibodies have been associated with protection against EBOV infection. In order to elicit protective neutralizing antibodies through vaccination a native-like conformation of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service